Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01382470_m1
          See other ZBED5-AS1 GE Assays ›
          Gene Symbol:
          ZBED5-AS1
          Gene Name:
          ZBED5 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 10858217 - 10879276 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01382470_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ZBED5-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_034137.1 2 - 3 219 67
          GenBank mRNA BC014360.1 - 2 - 3 181 67
          BC036635.1 - 2 - 3 162 67
          BG618678.1 - 2 - 3 220 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100012963.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000190.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28037211 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07637711 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07637745 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28037216 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28037213 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3336422 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 21926 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P39265688 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3336422 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3336422 - -
          Agilent Human 1B Microarray G4111A A_32_P446374 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 235010_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR11002561.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11002561.hg.1 NR_034137.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11002561.hg.1 TC11000190.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100012963.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012963.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012963.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012963.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012963.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012963.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012963.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012963.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012963.hg.1 ENST00000529014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012963.hg.1 ENST00000529014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012963.hg.1 ENST00000529014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012963.hg.1 ENST00000529014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012963.hg.1 ENST00000529014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012963.hg.1 ENST00000529014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012963.hg.1 ENST00000529014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012963.hg.1 ENST00000529014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012963.hg.1 NR_034137.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012963.hg.1 NR_034137.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012963.hg.1 NR_034137.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012963.hg.1 NR_034137.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012963.hg.1 NR_034137.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100199734.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100199734.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100199734.hg.1 NR_034137.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100199734.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100199734.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100199734.hg.1 NR_034137.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100199734.hg.1 TC1100012963.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100199733.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100199733.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100199733.hg.1 ENST00000529014.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100199733.hg.1 ENST00000529014.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100199733.hg.1 NR_034137.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100199733.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100199733.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100199733.hg.1 ENST00000529014.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100199733.hg.1 ENST00000529014.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100199733.hg.1 NR_034137.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100199733.hg.1 TC1100012963.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100012963.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012963.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012963.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012963.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012963.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012963.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012963.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012963.hg.1 ENST00000501079.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012963.hg.1 ENST00000529014.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012963.hg.1 ENST00000529014.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012963.hg.1 ENST00000529014.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012963.hg.1 ENST00000529014.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012963.hg.1 ENST00000529014.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012963.hg.1 ENST00000529014.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012963.hg.1 ENST00000529014.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012963.hg.1 ENST00000529014.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012963.hg.1 NR_034137.1 Refseq
          Affymetrix Human Clariom D Assay TC1100012963.hg.1 NR_034137.1 Refseq
          Affymetrix Human Clariom D Assay TC1100012963.hg.1 NR_034137.1 Refseq
          Affymetrix Human Clariom D Assay TC1100012963.hg.1 NR_034137.1 Refseq
          Affymetrix Human Clariom D Assay TC1100012963.hg.1 NR_034137.1 Refseq
          Human Genome U133 Plus 2.0 Array 235010_PM_at - -
          Human Genome U133 Plus 2.0 Array 235010_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235010_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235010_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235010_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235010_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235010_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235010_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235010_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235010_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235010_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235010_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235010_at3 - -
          Human Genome U95 Set 51234_PM_at - -
          Human Genome U95 Set 51234_at - -
          Human Genome U133 Set 235010_PM_at - -
          Human Genome U133 Set 235010_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046148 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046148 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 115081 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline