Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01387844_m1
          See other ZNF717 GE Assays ›
          Gene Symbol:
          ZNF717
          Gene Name:
          zinc finger protein 717
          Gene Aliases:
          OB1, X17, ZNF838
          Chromosome Location:
          Chr.3: 75694701 - 75785549 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01387844_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ZNF717
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001128223.3 NP_001121695.1 3 - 4 400 74
          NM_001290208.3 NP_001277137.1 3 - 4 347 74
          NM_001290209.3 NP_001277138.1 3 - 4 376 74
          NM_001290210.2 NP_001277139.1 3 - 4 347 74
          NM_001324026.2 NP_001310955.1 4 - 5 566 74
          NM_001324027.1 NP_001310956.1 3 - 4 400 74
          NM_001324028.1 NP_001310957.1 3 - 4 531 74
          XM_005264711.5 XP_005264768.1 4 - 5 449 74
          XM_011533244.4 XP_011531546.1 4 - 5 313 74
          XM_011533245.4 XP_011531547.1 4 - 5 435 74
          XM_011533246.4 XP_011531548.1 4 - 5 313 74
          XM_011533248.3 XP_011531550.1 4 - 5 313 74
          XM_011533249.4 XP_011531551.1 4 - 5 313 74
          XM_011533250.4 XP_011531552.1 4 - 5 313 74
          XM_011533251.2 XP_011531553.1 4 - 5 313 74
          XM_024453284.2 XP_024309052.1 3 - 4 531 74
          XM_047447038.1 XP_047302994.1 4 - 5 327 74
          XM_047447039.1 XP_047302995.1 3 - 4 225 74
          XM_047447040.1 XP_047302996.1 4 - 5 313 74
          XM_047447041.1 XP_047302997.1 4 - 5 313 74
          GenBank mRNA AK123776.1 - 3 - 4 446 74
          BQ928230.1 - 3 - 4 74
          DA618280.1 - 3 - 4 450 74
          DB079386.1 - 3 - 4 74
          DR006008.1 - 3 - 4 456 74
          HY057162.1 - 3 - 4 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300011597.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03001565.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_001290209.3 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          negative regulation of transcription, DNA-templated
          DNA binding
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100017566.hg.1 ENST00000412638.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017566.hg.1 ENST00000412638.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017566.hg.1 ENST00000412638.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017566.hg.1 ENST00000412638.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017566.hg.1 ENST00000412638.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017566.hg.1 ENST00000412638.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017566.hg.1 ENST00000412638.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017566.hg.1 ENST00000412638.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03028446.hg.1 ENST00000400845.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03028446.hg.1 ENST00000422325.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03028446.hg.1 ENST00000477374.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03028446.hg.1 ENST00000478296.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03028446.hg.1 ENST00000491507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03028446.hg.1 NM_001128223.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03028446.hg.1 TC03001565.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000012285.hg.1 ENST00000620178.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012285.hg.1 ENST00000620178.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012285.hg.1 ENST00000620178.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000468296.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000468296.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000468296.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000468296.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000468296.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000468296.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000468296.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000471541.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000471541.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000471541.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000471541.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000471541.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000471541.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000477374.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000477374.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000477374.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000477374.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000477374.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000477374.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000477374.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000478296.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000478296.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000478296.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000478296.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000478296.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000478296.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000478296.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000491507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000491507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000491507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000491507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000491507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000491507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 ENST00000491507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001128223.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001128223.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001128223.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001128223.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001128223.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001128223.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001128223.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001128223.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001128223.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001290208.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001290208.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001290208.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001290208.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001290208.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001290208.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001290208.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001290209.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001290209.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001290209.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001290209.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001290209.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001290209.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001290209.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001290210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001290210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001290210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001290210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001290210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001290210.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011597.hg.1 NM_001290210.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012285.hg.1 ENST00000620178.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012285.hg.1 ENST00000620178.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012285.hg.1 ENST00000620178.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000468296.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000468296.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000468296.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000468296.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000468296.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000468296.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000468296.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000471541.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000471541.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000471541.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000471541.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000471541.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000471541.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000477374.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000477374.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000477374.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000477374.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000477374.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000477374.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000477374.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000478296.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000478296.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000478296.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000478296.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000478296.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000478296.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000478296.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000491507.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000491507.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000491507.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000491507.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000491507.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000491507.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 ENST00000491507.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001128223.2 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001128223.2 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001128223.2 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001128223.2 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001128223.2 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001128223.2 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001128223.2 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001128223.2 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001128223.2 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001290208.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001290208.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001290208.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001290208.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001290208.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001290208.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001290208.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001290209.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001290209.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001290209.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001290209.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001290209.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001290209.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001290209.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001290210.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001290210.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001290210.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001290210.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001290210.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001290210.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011597.hg.1 NM_001290210.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017566.hg.1 ENST00000412638.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100017566.hg.1 ENST00000412638.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100017566.hg.1 ENST00000412638.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100017566.hg.1 ENST00000412638.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100017566.hg.1 ENST00000412638.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100017566.hg.1 ENST00000412638.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100017566.hg.1 ENST00000412638.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100017566.hg.1 ENST00000412638.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186620.hg.1 ENST00000468296.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186620.hg.1 ENST00000471541.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186620.hg.1 ENST00000477374.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186620.hg.1 ENST00000478296.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186620.hg.1 ENST00000491507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186620.hg.1 NM_001128223.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300186620.hg.1 NM_001290208.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300186620.hg.1 NM_001290209.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300186620.hg.1 NM_001290210.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300186620.hg.1 ENST00000468296.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186620.hg.1 ENST00000471541.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186620.hg.1 ENST00000477374.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186620.hg.1 ENST00000478296.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186620.hg.1 ENST00000491507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300186620.hg.1 NM_001128223.2 Refseq
          Affymetrix Human Clariom D Assay PSR0300186620.hg.1 NM_001290208.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300186620.hg.1 NM_001290209.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300186620.hg.1 NM_001290210.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300186620.hg.1 TC0300011597.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline