Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01391389_m1
          See other LOC284009 GE Assays ›
          Gene Symbol:
          LOC284009
          Gene Name:
          uncharacterized LOC284009
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 2406981 - 2415436 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01391389_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC284009
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028335.1 2 - 3 328 60
          GenBank mRNA AK093990.1 - 2 - 3 975 60
          BC093711.1 - 2 - 3 329 7939556 60
          BC112034.1 - 2 - 3 329 8327502 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700012328.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000995.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3269864 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3269869 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 62621 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 41213 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3269864 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3269864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10194218 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10194223 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10194221 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10194214 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10194192 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30743235 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30743239 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10194194 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30743268 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10194189 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10194195 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10194227 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30743232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30743270 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30743256 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10194216 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10194223 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30743235 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10194216 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30743239 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30743256 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10194214 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30743268 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10194195 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30743270 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10194227 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10194189 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10194218 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10194194 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10194192 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30743232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10194221 - -
          Agilent Human 1B Microarray G4111A A_32_P939292 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8011271 - -
          Affymetrix Human Gene 1.0 ST Array 8011274 - -
          Affymetrix Human Gene 1.0 ST Array 8011273 - -
          Affymetrix Human Gene 1.0 ST Array 8011272 - -
          Affymetrix Human Gene 1.1 ST Array 8011270 - -
          Affymetrix HT HG-U133+ PM Array Plate 1557399_PM_at - -
          Affymetrix Human Genome U219 Array 11735649_PM_at - -
          Affymetrix Human Genome U219 Array 11735649_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1700012328.hg.1 ENST00000381977.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012328.hg.1 ENST00000381977.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012328.hg.1 ENST00000381977.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012328.hg.1 ENST00000381977.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012328.hg.1 NR_028335.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012328.hg.1 NR_028335.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012328.hg.1 NR_028335.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012328.hg.1 NR_028335.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012328.hg.1 NR_028335.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17013279.hg.1 ENST00000381977.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17013279.hg.1 NR_028335.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17013279.hg.1 TC17000995.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1700204514.hg.1 NR_028335.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700204514.hg.1 TC1700012328.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700204514.hg.1 NR_028335.1 Refseq
          Affymetrix Human Clariom D Assay TC1700012328.hg.1 ENST00000381977.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700012328.hg.1 ENST00000381977.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700012328.hg.1 ENST00000381977.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700012328.hg.1 ENST00000381977.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700012328.hg.1 NR_028335.1 Refseq
          Affymetrix Human Clariom D Assay TC1700012328.hg.1 NR_028335.1 Refseq
          Affymetrix Human Clariom D Assay TC1700012328.hg.1 NR_028335.1 Refseq
          Affymetrix Human Clariom D Assay TC1700012328.hg.1 NR_028335.1 Refseq
          Affymetrix Human Clariom D Assay TC1700012328.hg.1 NR_028335.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700204515.hg.1 ENST00000381977.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700204515.hg.1 NR_028335.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700204515.hg.1 TC1700012328.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700204515.hg.1 ENST00000381977.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700204515.hg.1 NR_028335.1 Refseq
          Human Genome U133 Plus 2.0 Array 238931_PM_at - -
          Human Genome U133 Plus 2.0 Array 1557399_at - -
          Human Genome U133 Plus 2.0 Array 238931_at - -
          Human Genome U133 Plus 2.0 Array 1557399_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557399_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557399_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557399_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557399_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557399_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557399_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557399_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557399_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557399_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557399_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557399_at4 - -
          Human Genome U95 Set 72242_PM_at - -
          Human Genome U95 Set 72242_at - -
          Human Genome U95 Set 80232_at - -
          Human Genome U95 Set 80232_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A055001 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A055001 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1790896 AK093990.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1790896 BC093711.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1790896 BC112034.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1790896 NR_028335.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1743532 - -
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1790896 AK093990.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1790896 BC093711.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1790896 BC112034.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1790896 NR_028335.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1743532 - -
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1743532 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1790896 AK093990.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1790896 BC093711.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1790896 BC112034.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1790896 NR_028335.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1790896 AK093990.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1790896 BC093711.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1790896 BC112034.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1790896 NR_028335.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1790896 AK093990.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1790896 BC093711.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1790896 BC112034.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1790896 NR_028335.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1790896 AK093990.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1790896 BC093711.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1790896 BC112034.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1790896 NR_028335.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline