Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01549924_g1
          See other LINC00239 GE Assays ›
          Gene Symbol:
          LINC00239
          Gene Name:
          long intergenic non-protein coding RNA 239
          Gene Aliases:
          C14orf72, NCRNA00239
          Chromosome Location:
          Chr.14: 101730437 - 101732525 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01549924_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00239
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026774.1 3 - 4 353 83
          GenBank mRNA BX113211.1 - 3 - 4 353 83
          CK904714.1 - 1 - 2 95 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400008332.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14000772.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P176911 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4768 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4769 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 12521 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 28185 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P176911 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P176911 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09520530 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09520542 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09520536 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29983037 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29983040 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29983043 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09520535 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09520540 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29983041 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09520527 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29983046 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29983048 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29983036 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09520536 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29983043 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09520542 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29983036 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09520527 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29983040 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29983041 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09520530 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29983037 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09520535 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29983048 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29983046 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09520540 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P176911 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520535 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520529 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520579 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520525 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520553 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520555 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520551 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520539 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520558 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520574 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520563 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520580 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520582 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16735658 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520546 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520548 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520532 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520540 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520527 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520568 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520543 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520561 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520571 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520573 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520560 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520566 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09520536 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1400008332.hg.1 ENST00000556973.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008332.hg.1 ENST00000556973.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008332.hg.1 ENST00000556973.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008332.hg.1 ENST00000556973.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008332.hg.1 ENST00000556973.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008332.hg.1 NR_026774.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008332.hg.1 NR_026774.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008332.hg.1 NR_026774.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008332.hg.1 NR_026774.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008332.hg.1 NR_026774.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14008261.hg.1 ENST00000556973.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14008261.hg.1 NR_026774.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14008261.hg.1 TC14000772.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1400008332.hg.1 ENST00000556973.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008332.hg.1 ENST00000556973.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008332.hg.1 ENST00000556973.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008332.hg.1 ENST00000556973.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008332.hg.1 ENST00000556973.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008332.hg.1 NR_026774.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008332.hg.1 NR_026774.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008332.hg.1 NR_026774.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008332.hg.1 NR_026774.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008332.hg.1 NR_026774.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400159239.hg.1 ENST00000556973.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400159239.hg.1 NR_026774.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400159239.hg.1 ENST00000556973.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400159239.hg.1 NR_026774.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400159239.hg.1 TC1400008332.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A051799 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A051799 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 160535 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1658628 NR_026774.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1658628 AI304659.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1658628 BX113211.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1658628 CK904714.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1658628 NR_026774.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1658628 AI304659.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1658628 BX113211.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1658628 CK904714.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1658628 NR_026774.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1658628 AI304659.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1658628 BX113211.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1658628 CK904714.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1658628 NR_026774.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1658628 AI304659.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1658628 BX113211.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1658628 CK904714.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline