Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01554513_g1
          See other MAD2L1 GE Assays ›
          Gene Symbol:
          MAD2L1
          Gene Name:
          MAD2 mitotic arrest deficient-like 1 (yeast)
          Gene Aliases:
          HSMAD2, MAD2
          Chromosome Location:
          Chr.4: 120059422 - 120066858 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01554513_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MAD2L1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002358.3 NP_002349.1 1 - 2 195 65
          GenBank mRNA AF394735.1 - 1 - 2 88 65
          AJ000186.1 - 1 - 2 137 65
          AK298228.1 - 1 - 2 170 65
          AK313827.1 - 1 - 2 157 65
          BC000356.2 - 1 - 2 145 2964388 65
          BC005945.1 - 1 - 2 138 4132892 65
          BC070283.1 - 1 - 2 107 6654207 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400011721.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04001507.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MAD2L1

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic sister chromatid segregation
          sister chromatid cohesion
          mitotic cell cycle checkpoint
          mitotic spindle assembly checkpoint
          anaphase-promoting complex-dependent catabolic process
          negative regulation of protein catabolic process
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of apoptotic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          negative regulation of mitotic cell cycle
          cell division
          negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
          positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
          positive regulation of mitotic cell cycle spindle assembly checkpoint
          negative regulation of ubiquitin protein ligase activity
          protein binding
          identical protein binding
          protein homodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR04021357.hg.1 TC04001507.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04021357.hg.1 ENST00000296509.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04021357.hg.1 NM_002358.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000511295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000511295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000511295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000511295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000511295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000512484.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000512484.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000512484.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 ENST00000512484.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 NM_002358.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 NM_002358.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 NM_002358.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 NM_002358.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 NM_002358.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 NM_002358.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 NM_002358.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 NM_002358.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 NM_002358.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011721.hg.1 NM_002358.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400007370.hg.1 ENST00000556153.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04021356.hg.1 TC04001507.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04021356.hg.1 ENST00000296509.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04021356.hg.1 NM_002358.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04021358.hg.1 TC04001507.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04021358.hg.1 ENST00000296509.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04021358.hg.1 NM_002358.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007761.hg.1 ENST00000587471.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180575.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180575.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180575.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180575.hg.1 ENST00000511295.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180575.hg.1 NM_002358.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400180575.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180575.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180575.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180575.hg.1 ENST00000511295.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180575.hg.1 NM_002358.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400180575.hg.1 TC0400011721.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400180577.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180577.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180577.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180577.hg.1 ENST00000511295.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180577.hg.1 NM_002358.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400180577.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180577.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180577.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180577.hg.1 ENST00000511295.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180577.hg.1 NM_002358.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400180577.hg.1 TC0400011721.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400180574.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180574.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180574.hg.1 ENST00000511295.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180574.hg.1 NM_002358.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400180574.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180574.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180574.hg.1 ENST00000511295.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180574.hg.1 NM_002358.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400180574.hg.1 TC0400011721.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000511295.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000511295.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000511295.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000511295.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000511295.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000512484.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000512484.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000512484.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 ENST00000512484.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 NM_002358.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 NM_002358.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 NM_002358.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 NM_002358.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 NM_002358.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 NM_002358.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 NM_002358.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 NM_002358.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 NM_002358.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011721.hg.1 NM_002358.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007761.hg.1 ENST00000587471.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180576.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180576.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180576.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180576.hg.1 ENST00000511295.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180576.hg.1 NM_002358.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400180576.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180576.hg.1 ENST00000333047.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180576.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180576.hg.1 ENST00000511295.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180576.hg.1 NM_002358.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400180576.hg.1 TC0400011721.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400180581.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180581.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180581.hg.1 NM_002358.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400180581.hg.1 ENST00000296509.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180581.hg.1 ENST00000504707.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180581.hg.1 NM_002358.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400180581.hg.1 TC0400011721.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1400007370.hg.1 ENST00000556153.3 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline