Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01560359_m1
          See other ZNF197 GE Assays ›
          Gene Symbol:
          ZNF197 ZNF660-ZNF197
          Gene Name:
          zinc finger protein 197
          ZNF660-ZNF197 readthrough
          Gene Aliases:
          D3S1363E, P18, VHLaK, ZKSCAN9, ZNF166, ZNF20, ZSCAN41
          -
          Chromosome Location:
          Chr.3: 150390 - 173825 on Build GRCh38
          Chromosome Location:
          Chr.3: 110318 - 173825 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs01560359_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ZNF197
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024855.3 NP_001020026.1 3 - 4 750 64
          NM_001323293.2 NP_001310222.1 3 - 4 750 64
          NM_001323294.2 NP_001310223.1 3 - 4 741 64
          NM_001323295.2 NP_001310224.1 4 - 5 874 64
          NM_006991.5 NP_008922.1 3 - 4 741 64
          NR_136582.2 4 - 5 874 64
          GenBank mRNA AF011573.2 - 2 - 3 556 64
          AK314196.1 - 3 - 4 737 64
          AY074878.1 - 3 - 4 737 64
          AY261677.1 - 2 - 3 570 64
          AY279353.1 - 3 - 4 758 64
          BC017584.1 - 2 - 3 722 64
          Z21707.1 - 2 - 3 468 64
          Gene Symbol: ZNF660-ZNF197
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001351732.2 NP_001338661.1 4 - 5 786 64
          NM_001351733.2 NP_001338662.1 4 - 5 795 64
          NM_001351734.2 NP_001338663.1 4 - 5 786 64
          NM_001351735.2 NP_001338664.1 4 - 5 795 64
          NR_147691.2 5 - 6 919 64
          NR_147692.2 5 - 6 919 64

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the NM_001323295.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ZNF197
          Click here to find siRNAs for Gene ZNF660-ZNF197

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          DNA binding
          zinc ion binding
          metal ion binding
          sequence-specific double-stranded DNA binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000344387.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000344387.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000344387.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000344387.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000344387.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000344387.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000344387.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000344387.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000344387.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000344387.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000396058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000396058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000396058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000396058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000396058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000396058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000396058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000396058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000396058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000396058.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 ENST00000625541.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_006991.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_006991.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_006991.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_006991.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_006991.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_006991.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_006991.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_006991.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_006991.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_006991.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013821.hg.1 NM_006991.4 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000334075.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000344387.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000344387.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000344387.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000344387.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000344387.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000344387.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000344387.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000344387.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000344387.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000344387.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383744.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000383745.6 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000396058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000396058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000396058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000396058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000396058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000396058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000396058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000396058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000396058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000396058.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 ENST00000625541.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_001024855.2 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_006991.4 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_006991.4 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_006991.4 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_006991.4 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_006991.4 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_006991.4 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_006991.4 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_006991.4 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_006991.4 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_006991.4 Refseq
          Affymetrix Human Clariom D Assay TC0300013821.hg.1 NM_006991.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 207006 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline