Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01568973_m1
          See other LINC00334 GE Assays ›
          Gene Symbol:
          LINC00334
          Gene Name:
          long intergenic non-protein coding RNA 334
          Gene Aliases:
          C21orf89, NCRNA00334
          Chromosome Location:
          Chr.21: 45234340 - 45258730 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01568973_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00334
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_135279.1 2 - 3 252 79
          GenBank mRNA AF426268.1 - 2 - 3 240 79

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AF426268.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17243331 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415922 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32078769 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415923 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415926 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415920 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32078746 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32078757 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415921 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415942 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415914 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415930 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415948 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415915 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415946 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415917 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415910 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415909 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17243332 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415933 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17243335 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415955 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415954 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415944 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32078758 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415936 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32078766 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32078744 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32078725 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32078731 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415951 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415928 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415947 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32078730 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32078749 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415945 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32078726 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415937 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415952 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415935 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11415953 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P319557 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P319557 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 6160 - -
          Agilent Human Genome CGH Microarray 44B A_14_P130662 - -
          Agilent Human Genome CGH Microarray 44A A_14_P130662 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 32648 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P319557 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P319557 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P130662 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P03596473 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P41467729 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P130662 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03596462 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03596473 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03596455 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P319557 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 99371 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 49396 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 46471 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 64613 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03596451 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P22597570 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03596469 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03596455 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03596473 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P130662 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P41467729 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03596462 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P13941812 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21269431 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21269407 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03596465 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array XR_040130_at - -
          Affymetrix Human Gene 1.0 ST Array XR_040132_at - -
          Affymetrix Human Gene 1.0 ST Array XR_040131_at - -
          Affymetrix Human Gene 1.0 ST Array 8069204 - -
          Affymetrix Human Gene 1.0 ST Array 8069202 - -
          Affymetrix Human Gene 1.0 ST Array 8069203 - -
          Affymetrix Human Gene 1.0 ST Array 8069205 - -
          Affymetrix Human Gene 1.0 ST Array 8069201 - -
          Affymetrix Human Gene 1.1 ST Array 8069200 - -
          Affymetrix HT HG-U133+ PM Array Plate 240009_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1552876_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1552877_PM_s_at - -
          Affymetrix Human Genome U219 Array 11761892_PM_at - -
          Affymetrix Human Genome U219 Array 11761892_at - -
          Affymetrix Human Transcriptome Array 2.0 TC2100007406.hg.1 ENST00000328344.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007406.hg.1 ENST00000328344.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007406.hg.1 ENST00000328344.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007406.hg.1 ENST00000328344.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007406.hg.1 ENST00000584169.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007406.hg.1 ENST00000584169.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007406.hg.1 ENST00000584169.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007406.hg.1 ENST00000584169.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007406.hg.1 ENST00000584169.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007406.hg.1 ENST00000584169.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007406.hg.1 ENST00000328344.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100007406.hg.1 ENST00000328344.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100007406.hg.1 ENST00000328344.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100007406.hg.1 ENST00000328344.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100007406.hg.1 ENST00000584169.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007406.hg.1 ENST00000584169.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007406.hg.1 ENST00000584169.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007406.hg.1 ENST00000584169.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007406.hg.1 ENST00000584169.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007406.hg.1 ENST00000584169.1 Ensembl
          Human Genome U133 Plus 2.0 Array 240009_PM_at - -
          Human Genome U133 Plus 2.0 Array 1552877_s_at - -
          Human Genome U133 Plus 2.0 Array 1552876_at - -
          Human Genome U133 Plus 2.0 Array 240009_at - -
          Human Genome U133 Plus 2.0 Array 1552876_PM_at - -
          Human Genome U133 Plus 2.0 Array 1552877_PM_s_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552877_s_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240009_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552877_s_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552876_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240009_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552876_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552876_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552877_s_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552877_s_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552876_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240009_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552876_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552877_s_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552877_s_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552877_s_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552876_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552876_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240009_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240009_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240009_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552876_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240009_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552876_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240009_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240009_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552877_s_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240009_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552877_s_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552877_s_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552876_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240009_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552877_s_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552876_at11 - -
          Human Genome U95 Set 61544_at - -
          Human Genome U95 Set 53959_at - -
          Human Genome U95 Set 53959_PM_at - -
          Human Genome U95 Set 61544_PM_at - -
          Human Genome U133 Set 240009_PM_at - -
          Human Genome U133 Set 240009_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 224935 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3263072 AF426268.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3263072 AF426268.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline