Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01583919_m1
          See other PRPF40B GE Assays ›
          Gene Symbol:
          PRPF40B
          Gene Name:
          pre-mRNA processing factor 40 homolog B
          Gene Aliases:
          HYPC
          Chromosome Location:
          Chr.12: 49622646 - 49644665 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01583919_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PRPF40B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001031698.3 NP_001026868.2 16 - 17 1588 118
          NM_001363607.2 NP_001350536.2 16 - 17 1588 118
          NM_001379030.1 NP_001365959.1 16 - 17 1558 118
          NM_001379031.1 NP_001365960.1 15 - 16 1507 118
          NM_001379032.1 NP_001365961.1 15 - 16 1507 118
          NM_001379033.1 NP_001365962.1 15 - 16 1477 118
          NM_001379034.1 NP_001365963.1 15 - 16 1477 118
          NM_001379035.1 NP_001365964.1 14 - 15 1396 118
          NM_001379036.1 NP_001365965.1 14 - 15 1396 118
          NM_001379037.1 NP_001365966.1 13 - 14 1231 118
          NM_012272.3 NP_036404.1 15 - 16 2084 118
          XM_006719324.5 XP_006719387.1 15 - 16 1867 118
          XM_006719325.5 XP_006719388.1 15 - 16 1867 118
          XM_011538139.4 XP_011536441.1 16 - 17 1588 118
          XM_011538140.4 XP_011536442.1 16 - 17 1588 118
          XM_011538143.4 XP_011536445.1 15 - 16 1507 118
          XM_017019135.3 XP_016874624.1 15 - 16 1492 118
          XM_017019138.2 XP_016874627.1 15 - 16 1872 118
          XM_047428674.1 XP_047284630.1 16 - 17 1607 118
          XM_047428675.1 XP_047284631.1 16 - 17 1591 118
          XM_047428676.1 XP_047284632.1 16 - 17 1607 118
          XM_047428677.1 XP_047284633.1 15 - 16 1507 118
          XM_047428678.1 XP_047284634.1 15 - 16 1563 118
          XM_047428679.1 XP_047284635.1 15 - 16 1873 118
          GenBank mRNA AL137459.1 - 2 - 3 238 118
          AL834216.1 - 15 - 16 2025 118
          BC031917.1 - 13 - 14 1331 118
          BC050398.2 - 16 - 17 1571 118
          BC067364.1 - 15 - 16 1556 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200012626.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12000377.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PRPF40B

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          mRNA processing
          RNA splicing
          mRNA cis splicing, via spliceosome
          RNA binding
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR12004680.hg.1 ENST00000261897.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12004680.hg.1 ENST00000380281.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12004680.hg.1 NM_001031698.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12004680.hg.1 NM_012272.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12004680.hg.1 TC12000377.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1200199855.hg.1 ENST00000261897.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200199855.hg.1 ENST00000380281.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200199855.hg.1 ENST00000548825.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200199855.hg.1 NM_001031698.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200199855.hg.1 NM_012272.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200199855.hg.1 ENST00000261897.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200199855.hg.1 ENST00000380281.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200199855.hg.1 ENST00000548825.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200199855.hg.1 NM_001031698.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200199855.hg.1 NM_012272.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200199855.hg.1 TC1200012626.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline