Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01598011_m1
          See other ZDHHC6 GE Assays ›
          Gene Symbol:
          ZDHHC6
          Gene Name:
          zinc finger DHHC-type containing 6
          Gene Aliases:
          DHHC-6, ZNF376
          Chromosome Location:
          Chr.10: 112424411 - 112448335 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01598011_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ZDHHC6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001303134.1 NP_001290063.1 7 - 8 1351 108
          NM_022494.2 NP_071939.1 7 - 8 1363 108
          XM_005270051.4 XP_005270108.1 7 - 8 2298 108
          XM_006717948.3 XP_006718011.1 7 - 8 1469 108
          XM_006717949.3 XP_006718012.1 6 - 7 1198 108
          XM_011540091.2 XP_011538393.1 7 - 8 1351 108
          XM_011540092.2 XP_011538394.1 7 - 8 1268 108
          XM_011540093.1 XP_011538395.1 7 - 8 1377 108
          XM_011540094.2 XP_011538396.1 7 - 8 1236 108
          XM_011540095.2 XP_011538397.1 6 - 7 1086 108
          XM_017016561.1 XP_016872050.1 7 - 8 1289 108
          XM_017016562.1 XP_016872051.1 7 - 8 1457 108
          XM_017016563.1 XP_016872052.1 7 - 8 1270 108
          XM_017016564.1 XP_016872053.1 7 - 8 1469 108
          XM_017016565.1 XP_016872054.1 7 - 8 1535 108
          XM_017016566.1 XP_016872055.1 7 - 8 1348 108
          XM_017016567.1 XP_016872056.1 6 - 7 1186 108
          XM_017016568.1 XP_016872057.1 7 - 8 1174 108
          XM_017016569.1 XP_016872058.1 3 - 4 311 108
          GenBank mRNA AF267740.1 - 7 - 8 1317 108
          AK025605.1 - 7 - 8 1318 108
          AK026369.1 - 2 - 3 223 108
          AK223086.1 - 7 - 8 1317 108
          BC007213.1 - 7 - 8 1293 2960984 108
          BC017434.2 - 7 - 8 1293 2960984 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000012595.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10001667.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ZDHHC6

          Gene Ontology Categories:

          Function(s) Process(es)

          protein palmitoylation
          zinc ion binding
          palmitoyltransferase activity
          protein-cysteine S-palmitoyltransferase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000471035.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000471035.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000471035.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000471035.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000471035.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000482410.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000482410.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000482410.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000482410.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 ENST00000482410.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10021862.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10021862.hg.1 ENST00000369405.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10021862.hg.1 ENST00000482410.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10021862.hg.1 NM_022494.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10021862.hg.1 TC10001667.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10021863.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10021863.hg.1 ENST00000369405.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10021863.hg.1 ENST00000482410.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10021863.hg.1 NM_022494.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10021863.hg.1 TC10001667.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000471035.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000471035.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000471035.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000471035.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000471035.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000482410.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000482410.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000482410.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000482410.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 ENST00000482410.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012595.hg.1 NM_022494.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000188022.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000188022.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000188022.hg.1 ENST00000471035.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000188022.hg.1 ENST00000482410.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000188022.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000188022.hg.1 NM_022494.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000188022.hg.1 ENST00000369404.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000188022.hg.1 ENST00000369405.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000188022.hg.1 ENST00000471035.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000188022.hg.1 ENST00000482410.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000188022.hg.1 NM_001303134.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000188022.hg.1 NM_022494.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000188022.hg.1 TC1000012595.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline