Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01686818_s1
          See other OR4C12 GE Assays ›
          Gene Symbol:
          OR4C12
          Gene Name:
          olfactory receptor family 4 subfamily C member 12
          Gene Aliases:
          OR11-259
          Chromosome Location:
          Chr.11: 49981473 - 49982535 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01686818_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OR4C12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005270.4 NP_001005270.3 1 717 139
          GenBank mRNA BC136850.1 - 1 717 9020841 139
          BC136851.1 - 1 717 9020842 139

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100010809.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11001760.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene OR4C12

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          G-protein coupled receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16449881 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28212103 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07804964 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28212056 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28212052 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07804963 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28212104 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07804961 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28212083 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16449822 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16449837 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28212064 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28212072 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28212089 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07804962 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28212045 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16449812 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16449855 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 962131 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3272070 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0603 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0602 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 39944 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P11094146 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3272070 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3272070 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212064 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212067 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212083 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07804962 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07804967 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07804964 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212099 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16449837 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07804966 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07804969 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212089 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212104 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07804968 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212103 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212066 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16449886 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212072 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07804963 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16449881 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212067 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212104 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07804967 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07804963 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07804968 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212066 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212083 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07804962 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07804966 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16449837 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212103 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212072 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212099 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16449881 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07804964 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07804969 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212064 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16449886 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28212089 - -
          Agilent Human 1B Microarray G4111A A_32_P724397 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07804969 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07804963 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07804960 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07804967 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07804965 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16449809 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07804962 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001005270_at - -
          Affymetrix Human Gene 1.0 ST Array 7948091 - -
          Affymetrix Human Gene 1.0 ST Array 7948104 - -
          Affymetrix Human Gene 1.1 ST Array 7948103 - -
          Affymetrix Human Gene 1.1 ST Array 7948090 - -
          Affymetrix Human Genome U219 Array 11738717_at - -
          Affymetrix Human Genome U219 Array 11738717_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1100010809.hg.1 ENST00000335238.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010809.hg.1 ENST00000335238.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010809.hg.1 ENST00000335238.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010809.hg.1 NM_001005270.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010809.hg.1 NM_001005270.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010809.hg.1 NM_001005270.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007520.hg.1 ENST00000530855.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007567.hg.1 ENST00000330155.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11020637.hg.1 ENST00000335238.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11020637.hg.1 NM_001005270.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11020637.hg.1 TC11001760.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1100007567.hg.1 ENST00000330155.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100010809.hg.1 ENST00000335238.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100010809.hg.1 ENST00000335238.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100010809.hg.1 ENST00000335238.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100010809.hg.1 NM_001005270.4 Refseq
          Affymetrix Human Clariom D Assay TC1100010809.hg.1 NM_001005270.4 Refseq
          Affymetrix Human Clariom D Assay TC1100010809.hg.1 NM_001005270.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100181970.hg.1 ENST00000335238.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100181970.hg.1 NM_001005270.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100181970.hg.1 ENST00000335238.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100181970.hg.1 NM_001005270.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100181970.hg.1 TC1100010809.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100181969.hg.1 ENST00000335238.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100181969.hg.1 NM_001005270.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100181969.hg.1 ENST00000335238.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100181969.hg.1 NM_001005270.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100181969.hg.1 TC1100010809.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100181968.hg.1 ENST00000335238.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100181968.hg.1 NM_001005270.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100181968.hg.1 ENST00000335238.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100181968.hg.1 NM_001005270.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100181968.hg.1 TC1100010809.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100007520.hg.1 ENST00000530855.1 Ensembl
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045143 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045143 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 210738 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2066904 NM_001005270.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2066904 BC136850.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2066904 BC136851.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1777706 NM_001005270.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1777706 BC136850.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1777706 BC136851.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1777706 NM_001005270.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1777706 BC136850.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1777706 BC136851.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2066904 NM_001005270.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2066904 BC136850.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2066904 BC136851.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1777706 NM_001005270.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1777706 BC136850.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1777706 BC136851.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2066904 NM_001005270.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2066904 BC136850.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2066904 BC136851.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1777706 NM_001005270.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1777706 BC136850.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1777706 BC136851.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1777706 NM_001005270.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1777706 BC136850.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1777706 BC136851.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2066904 NM_001005270.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2066904 BC136850.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2066904 BC136851.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_18993-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1777706 NM_001005270.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1777706 BC136850.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1777706 BC136851.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1777706 NM_001005270.2 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1777706 BC136850.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1777706 BC136851.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 2940273 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_18993 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2066904 NM_001005270.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2066904 BC136850.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2066904 BC136851.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1777706 NM_001005270.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1777706 BC136850.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1777706 BC136851.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2066904 NM_001005270.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2066904 BC136850.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2066904 BC136851.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1777706 NM_001005270.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1777706 BC136850.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1777706 BC136851.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2066904 NM_001005270.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2066904 BC136850.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2066904 BC136851.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1777706 NM_001005270.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1777706 BC136850.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1777706 BC136851.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2066904 NM_001005270.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2066904 BC136850.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2066904 BC136851.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline