Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01874526_u1
          See other CYP2A7 GE Assays ›
          Gene Symbol:
          CYP2A7
          Gene Name:
          cytochrome P450 family 2 subfamily A member 7
          Gene Aliases:
          CPA7, CPAD, CYP2A, CYPIIA7, P450-IIA4
          Chromosome Location:
          Chr.19: 40875439 - 40885952 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01874526_u1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CYP2A7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000764.2 NP_000755.2 2 748 117
          XM_017026383.1 XP_016881872.1 2 3945 117
          GenBank mRNA BC117165.1 - 2 251 40125716 117
          BC143325.1 - 2 251 9051830 117
          M33317.1 - 2 215 117
          U22029.1 - 2 749 117

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900011956.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19002702.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection P450 Recommended Assay Homo sapiens Inventoried Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CYP2A7

          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          epoxygenase P450 pathway
          oxidation-reduction process
          iron ion binding
          arachidonic acid epoxygenase activity
          steroid hydroxylase activity
          oxygen binding
          heme binding
          aromatase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19021116.hg.1 TC19002702.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19021116.hg.1 ENST00000301146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19021116.hg.1 NM_000764.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19021115.hg.1 TC19002702.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19021115.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19021115.hg.1 ENST00000301146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19021115.hg.1 NM_000764.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19021115.hg.1 NM_030589.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000594332.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000594332.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000594332.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000594332.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000598264.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000598264.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000598264.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000598264.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000598264.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000602008.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000602008.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000602008.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 ENST00000602008.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000594332.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000594332.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000594332.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000594332.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000598264.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000598264.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000598264.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000598264.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000598264.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000602008.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000602008.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000602008.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 ENST00000602008.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_000764.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Clariom D Assay TC1900011956.hg.1 NM_030589.2 Refseq
          Affymetrix Human Clariom D Assay PSR1900206311.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1900206311.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900206311.hg.1 ENST00000594332.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900206311.hg.1 NM_000764.2 Refseq
          Affymetrix Human Clariom D Assay PSR1900206311.hg.1 NM_030589.2 Refseq
          Affymetrix Human Clariom D Assay PSR1900206311.hg.1 TC1900011956.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900206311.hg.1 ENST00000291764.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1900206311.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900206311.hg.1 ENST00000594332.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900206311.hg.1 NM_000764.2 Refseq
          Affymetrix Human Clariom D Assay PSR1900206311.hg.1 NM_030589.2 Refseq
          Affymetrix Human Clariom D Assay PSR1900206312.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900206312.hg.1 NM_000764.2 Refseq
          Affymetrix Human Clariom D Assay PSR1900206312.hg.1 TC1900011956.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900206312.hg.1 ENST00000301146.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900206312.hg.1 NM_000764.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0