Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01943460_s1
          See other OR5B12 GE Assays ›
          Gene Symbol:
          OR5B12
          Gene Name:
          olfactory receptor family 5 subfamily B member 12
          Gene Aliases:
          OR11-241, OR5B12P, OR5B16, OST743
          Chromosome Location:
          Chr.11: 58438994 - 58442236 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01943460_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OR5B12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004733.3 NP_001004733.1 2 1046 142
          GenBank mRNA BC136947.1 - 1 858 142
          BC136948.1 - 1 858 142

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100010943.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11001809.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene OR5B12

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07817029 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07817030 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28226937 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07817052 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07817068 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07817054 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07817066 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16454778 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07817027 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28226948 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07817051 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07817036 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28226917 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28226919 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28226935 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07817031 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28226950 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07817059 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28226962 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28226941 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28226956 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07817043 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28226943 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28226942 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28226952 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07817026 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28226940 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28226946 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07817041 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07817037 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28226953 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07817032 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28226939 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28226930 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28226918 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07817069 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07817034 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07817024 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07817028 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28226955 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3380452 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0465 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_0466 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 54105 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P11111487 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3380452 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3380452 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226950 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226940 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226939 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07817051 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226954 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226956 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07817056 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226942 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226935 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226955 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07817032 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07817034 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226946 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07817043 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07817054 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16454778 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226958 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226953 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07817066 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226948 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07817069 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07817036 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07817059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226941 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226935 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16454778 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07817054 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226939 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226958 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226954 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07817069 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226942 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226950 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226946 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07817066 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07817036 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226948 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07817059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07817051 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226940 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226955 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07817034 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226956 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07817056 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226941 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28226953 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07817032 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07817043 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07817055 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07817069 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07817060 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07817066 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07817043 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07817049 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07817039 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07817046 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07817034 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07817036 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07817064 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07817032 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07817062 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07817051 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07817057 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07817063 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07817053 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P39368118 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P39368118 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817051 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817028 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817021 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817027 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817060 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817043 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16454778 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817062 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817069 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817039 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817034 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817019 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817053 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817025 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817030 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817064 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817049 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16454777 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817057 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817036 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817055 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817046 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817023 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07817032 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P39368118 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001004733_at - -
          Affymetrix Human Gene 1.0 ST Array 7948329 - -
          Affymetrix Human Gene 1.1 ST Array 7948328 - -
          Affymetrix Human Genome U219 Array 11738725_PM_at - -
          Affymetrix Human Genome U219 Array 11738725_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR11020923.hg.1 ENST00000302572.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11020923.hg.1 NM_001004733.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11020923.hg.1 TC11001809.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100010943.hg.1 ENST00000302572.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010943.hg.1 ENST00000302572.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010943.hg.1 NM_001004733.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010943.hg.1 NM_001004733.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100182593.hg.1 ENST00000302572.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100182593.hg.1 NM_001004733.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100182593.hg.1 ENST00000302572.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100182593.hg.1 NM_001004733.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100182593.hg.1 TC1100010943.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100010943.hg.1 ENST00000302572.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100010943.hg.1 ENST00000302572.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100010943.hg.1 NM_001004733.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010943.hg.1 NM_001004733.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100182592.hg.1 ENST00000302572.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100182592.hg.1 NM_001004733.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100182592.hg.1 ENST00000302572.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100182592.hg.1 NM_001004733.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100182592.hg.1 TC1100010943.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045211 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045211 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1740776 NM_001004733.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1740776 BC136947.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1740776 BC136948.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1740776 NM_001004733.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1740776 BC136947.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1740776 BC136948.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1740776 NM_001004733.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1740776 BC136947.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1740776 BC136948.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1740776 NM_001004733.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1740776 BC136947.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1740776 BC136948.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1740776 NM_001004733.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1740776 BC136947.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1740776 BC136948.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_21702-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1740776 NM_001004733.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1740776 BC136947.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1740776 BC136948.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1740776 NM_001004733.2 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1740776 BC136947.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1740776 BC136948.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 7330437 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_21702 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1740776 NM_001004733.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1740776 BC136947.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1740776 BC136948.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1740776 NM_001004733.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1740776 BC136947.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1740776 BC136948.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1740776 NM_001004733.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1740776 BC136947.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1740776 BC136948.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1740776 NM_001004733.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1740776 BC136947.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1740776 BC136948.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1740776 NM_001004733.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1740776 BC136947.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1740776 BC136948.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline