Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02576432_m1
          See other DEFB135 GE Assays ›
          Gene Symbol:
          DEFB135
          Gene Name:
          defensin beta 135
          Gene Aliases:
          DEFB136
          Chromosome Location:
          Chr.8: 11982321 - 11984590 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02576432_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DEFB135
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033017.2 NP_001028189.2 1 - 2 65 91
          GenBank mRNA AY621332.1 - 1 - 2 65 91
          DQ012025.1 - 1 - 2 65 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800006764.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08000095.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DEFB135

          Panther Classification:

          Molecular Function -

          antimicrobial response protein defense/immunity protein

          Gene Ontology Categories:

          Function(s)

          defense response to bacterium
          innate immune response

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239143 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959689 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239130 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959708 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959702 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959700 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959720 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239148 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239156 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959717 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959715 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239126 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239152 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959706 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959698 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239141 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239147 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959723 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959704 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239124 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959691 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959705 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239154 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239132 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239157 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959711 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239131 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26239144 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05959716 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 13035 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 19649 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 472510 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 371585 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3397545 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1189 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1190 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 2114 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P32207269 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P16387223 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P18217803 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3397545 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3397545 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239144 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959703 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239130 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959702 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959695 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239132 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959698 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959706 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959700 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959704 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239124 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959718 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959689 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959705 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239126 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959694 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959715 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239147 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239141 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959716 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959711 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959695 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959703 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239126 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239141 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959715 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959704 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959700 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239144 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959718 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239130 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959706 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959716 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959702 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239147 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959694 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959705 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959698 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959711 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05959689 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239124 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26239132 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P18217803 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P18217803 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 35839 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 104406 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01863386 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18217803 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16069035 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959669 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959735 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959744 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959695 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959700 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959697 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959676 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959725 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959663 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959688 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959667 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959672 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959733 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959690 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959717 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959728 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959692 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959711 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959713 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959707 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959679 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959724 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959702 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959705 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959703 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959674 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959709 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959731 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16069039 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959739 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959748 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959665 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959685 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959721 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959681 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959742 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959719 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05959737 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P18217803 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001033017_at - -
          Affymetrix Human Gene 1.0 ST Array 8144689 - -
          Affymetrix Human Gene 1.0 ST Array 8144690 - -
          Affymetrix Human Gene 1.0 ST Array 8144688 - -
          Affymetrix Human Gene 1.1 ST Array 8144687 - -
          Affymetrix Human Genome U219 Array 11738792_at - -
          Affymetrix Human Genome U219 Array 11738792_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR08000941.hg.1 TC08000095.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08000941.hg.1 ENST00000382208.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08000941.hg.1 NM_001033017.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08000942.hg.1 TC08000095.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08000942.hg.1 ENST00000382208.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08000942.hg.1 NM_001033017.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800006764.hg.1 ENST00000382208.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800006764.hg.1 ENST00000382208.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800006764.hg.1 NM_001033017.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800006764.hg.1 NM_001033017.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800145915.hg.1 ENST00000382208.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800145915.hg.1 NM_001033017.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800145915.hg.1 ENST00000382208.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800145915.hg.1 NM_001033017.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800145915.hg.1 TC0800006764.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800145914.hg.1 ENST00000382208.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800145914.hg.1 NM_001033017.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800145914.hg.1 ENST00000382208.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800145914.hg.1 NM_001033017.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800145914.hg.1 TC0800006764.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0800006764.hg.1 ENST00000382208.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800006764.hg.1 ENST00000382208.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800006764.hg.1 NM_001033017.2 Refseq
          Affymetrix Human Clariom D Assay TC0800006764.hg.1 NM_001033017.2 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A036359 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A036359 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1711790 NM_001033017.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1711790 AY621332.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1711790 DQ012025.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1711790 NM_001033017.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1711790 AY621332.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1711790 DQ012025.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1711790 NM_001033017.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1711790 AY621332.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1711790 DQ012025.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1711790 NM_001033017.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1711790 AY621332.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1711790 DQ012025.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1711790 NM_001033017.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1711790 AY621332.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1711790 DQ012025.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_12299-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1711790 NM_001033017.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1711790 AY621332.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1711790 DQ012025.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1711790 NM_001033017.2 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1711790 AY621332.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1711790 DQ012025.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 4040253 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_12299 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1711790 NM_001033017.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1711790 AY621332.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1711790 DQ012025.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1711790 NM_001033017.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1711790 AY621332.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1711790 DQ012025.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1711790 NM_001033017.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1711790 AY621332.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1711790 DQ012025.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1711790 NM_001033017.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1711790 AY621332.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1711790 DQ012025.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1711790 NM_001033017.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1711790 AY621332.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1711790 DQ012025.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline