Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03309977_s1
          See other PWAR1 GE Assays ›
          Gene Symbol:
          PWAR1
          Gene Name:
          Prader Willi/Angelman region RNA 1
          Gene Aliases:
          D15S227E, PAR-1, PAR1
          Chromosome Location:
          Chr.15: 25135642 - 25138053 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03309977_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PWAR1
          Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_022009.1
          1 2247 121
          GenBank mRNA AF019616.1 - LIT2055
          PAR1
          1 2251 121

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09544030 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09544009 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023856 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023875 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09544020 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09544035 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023859 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09544002 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023857 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09544007 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09544000 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023881 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023863 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023879 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023854 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09544037 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023872 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023858 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023874 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023877 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023868 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09544016 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023886 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09544001 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09544015 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09544036 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023867 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023850 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023851 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09544022 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09544019 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023873 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023885 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09544023 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023864 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023887 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09544005 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30023861 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 207791 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3259373 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4415 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4416 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 48370 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 20857 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3259373 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3259373 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544019 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023874 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544009 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023863 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544035 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544001 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544037 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023875 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023855 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023877 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544016 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023859 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544023 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023868 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544002 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544022 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023879 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544020 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023858 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023854 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023857 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544015 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023867 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023856 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544007 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544030 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023881 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023867 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023859 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544001 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544016 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544020 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023858 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544015 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023874 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544009 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544019 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544007 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544030 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023855 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544023 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544035 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023881 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544002 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023877 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023854 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023863 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023856 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023857 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544022 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023879 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09544037 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023868 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30023875 - -
          Agilent Human 1B Microarray G4111A A_32_P760922 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544000 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544017 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544071 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544036 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544056 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544038 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544070 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544023 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544055 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544033 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544005 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544065 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544046 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544018 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544026 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544059 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544034 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16760365 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544010 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544047 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544050 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544063 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544041 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544014 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544052 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544032 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544020 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544028 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544003 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544001 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544012 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544044 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09544008 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16760360 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052663 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052663 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3245815 AF019616.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3245815 NR_022009.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3245815 AF019616.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3245815 NR_022009.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3245815 AF019616.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3245815 NR_022009.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3245815 AF019616.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3245815 NR_022009.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline