Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03665449_s1
          See other SLC19A1 GE Assays ›
          Gene Symbol:
          SLC19A1
          Gene Name:
          solute carrier family 19 member 1
          Gene Aliases:
          CHMD, FOLT, IFC1, REFC, RFC1
          Chromosome Location:
          Chr.21: 45502517 - 45563143 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03665449_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 434.00
          Your Price
          Online offer: 407.65
          407.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLC19A1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_017028444.1 XP_016883933.1 7 2999 74
          XM_017028446.1 XP_016883935.1 6 2859 74
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB547170.1 - 1 166 Chr. 21 - 45513776 - 45514203 [-] on Build GRCh38 74
          GenBankmRNA BM016451.1 - 1 87 Chr. 21 - 45513205 - 45514123 [-] on Build GRCh38 74
          GenBankmRNA CA502870.1 - 1 Chr. 21 - 45513776 - 45514387 [+] on Build GRCh38 74
          GenBankmRNA GR939639.1 - 1 596 Chr. 21 - 45513852 - 45514632 [-] on Build GRCh38 74
          GenBankmRNA HY301483.1 - 1 Chr. 21 - 45513775 - 45514261 [+] on Build GRCh38 74
          GenBankmRNA HY216914.1 - 1 Chr. 21 - 45513775 - 45514266 [+] on Build GRCh38 74
          GenBankmRNA AI499793.1 - 1 Chr. 21 - 45513776 - 45514195 [+] on Build GRCh38 74
          GenBankmRNA DB540750.1 - 1 222 Chr. 21 - 45513776 - 45514258 [-] on Build GRCh38 74
          GenBankmRNA BF943560.1 - 1 Chr. 21 - 45513868 - 45514381 [+] on Build GRCh38 74
          GenBankmRNA GR939637.1 - 1 594 Chr. 21 - 45513867 - 45514621 [-] on Build GRCh38 74
          GenBankmRNA BU617054.1 - 1 Chr. 21 - 45513775 - 45514387 [+] on Build GRCh38 74
          GenBankmRNA BF312684.1 - 1 298 Chr. 21 - 45513807 - 45514334 [-] on Build GRCh38 74
          GenBankmRNA AA910025.1 - 1 Chr. 21 - 45513776 - 45514129 [+] on Build GRCh38 74
          GenBankmRNA AI204685.1 - 1 Chr. 21 - 45513776 - 45514176 [+] on Build GRCh38 74
          GenBankmRNA AL522263.3 - 1 651 Chr. 21 - 45513962 - 45514681 [-] on Build GRCh38 74
          GenBankmRNA BX389400.2 - 1 62 Chr. 21 - 45513376 - 45514081 [-] on Build GRCh38 74
          GenBankmRNA CA944419.1 - 1 Chr. 21 - 45513776 - 45514427 [+] on Build GRCh38 74
          GenBankmRNA BQ003811.1 - 1 Chr. 21 - 45513779 - 45514408 [+] on Build GRCh38 74
          GenBankmRNA GR939640.1 - 1 597 Chr. 21 - 45513874 - 45514635 [-] on Build GRCh38 74
          GenBankmRNA GR942535.1 - 1 Chr. 21 - 45513588 - 45514390 [+] on Build GRCh38 74
          GenBankmRNA BM011822.1 - 1 294 Chr. 21 - 45513483 - 45514329 [-] on Build GRCh38 74
          GenBankmRNA GR939635.1 - 1 596 Chr. 21 - 45513812 - 45514635 [-] on Build GRCh38 74
          GenBankmRNA GR939638.1 - 1 599 Chr. 21 - 45513857 - 45514637 [-] on Build GRCh38 74
          GenBankmRNA HY342873.1 - 1 Chr. 21 - 45513776 - 45514239 [+] on Build GRCh38 74

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream collagen type XVIII alpha 1 chain COL18A1 KNO,
          KNO1,
          KS
          -56.0 Anti-sense Chr. 21 - 45405137 - 45513720 on NCBI Build GRCh38
          Downstream uncharacterized LOC105372840 LOC105372840 76.6kb Anti-sense Chr. 21 - 45593664 - 45603056 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          female pregnancy
          folic acid transport
          folic acid metabolic process
          methotrexate transport
          reduced folate transmembrane transport
          folic acid binding
          folic acid transporter activity
          reduced folate carrier activity
          methotrexate transporter activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline