Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03680079_m1
          See other GNAS GE Assays ›
          Gene Symbol:
          GNAS
          Gene Name:
          GNAS complex locus
          Gene Aliases:
          AHO, C20orf45, GNAS1, GPSA, GSA, GSP, NESP, POH, SCG6, SgVI
          Chromosome Location:
          Chr.20: 58839681 - 58911196 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03680079_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GNAS
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000516.5 NP_000507.1 2 - 3 639 74
          NM_001077488.3 NP_001070956.1 2 - 3 639 74
          NM_001077490.2 NP_001070958.1 2 - 3 2429 74
          NM_001309840.1 NP_001296769.1 2 - 3 305 74
          NM_001309842.1 NP_001296771.1 2 - 3 639 74
          NM_001309861.1 NP_001296790.1 2 - 3 350 74
          NM_001309883.1 NP_001296812.1 4 - 5 2587 74
          NM_016592.3 NP_057676.1 2 - 3 1245 74
          NM_080425.3 NP_536350.2 2 - 3 2429 74
          NR_132272.1 2 - 3 122 74
          NR_132273.1 2 - 3 482 74
          XM_017027812.1 XP_016883301.1 2 - 3 2696 74
          XM_017027816.1 XP_016883305.1 2 - 3 845 74
          XM_017027821.1 XP_016883310.1 2 - 3 1282 74
          GenBank mRNA AF105253.1 - 2 - 3 1223 74
          AF493897.1 - 2 - 3 215 74
          AJ224867.1 - 4 - 5 610 74
          AK026564.1 - 3 - 4 409 74
          AK315860.1 - 2 - 3 137 74
          AK315874.1 - 2 - 3 318 74
          AL537118.3 - 2 - 3 335 73
          BC002722.2 - 3 - 4 333 74
          BC008855.2 - 3 - 4 357 74
          BM894355.1 - 2 - 3 180 74
          BT009905.1 - 2 - 3 215 74
          CN403086.1 - 2 - 3 263 74
          DB483472.1 - 2 - 3 218 74
          HY141824.1 - 2 - 3 122 74
          KU178095.1 - 2 - 3 215 74
          M14631.1 - 2 - 3 182 74
          X04408.1 - 2 - 3 283 74
          X07036.1 - 2 - 3 227 74
          X56009.1 - 2 - 3 291 74

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_017027816.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GNAS

          Panther Classification:

          Molecular Function -

          heterotrimeric G-protein

          Gene Ontology Categories:

          Function(s) Process(es)

          tissue homeostasis
          endochondral ossification
          energy reserve metabolic process
          DNA methylation
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          adenylate cyclase-activating dopamine receptor signaling pathway
          sensory perception of chemical stimulus
          embryonic hindlimb morphogenesis
          multicellular organism growth
          post-embryonic body morphogenesis
          response to drug
          positive regulation of osteoblast differentiation
          positive regulation of osteoclast differentiation
          developmental growth
          embryonic cranial skeleton morphogenesis
          cognition
          cartilage development
          bone development
          hair follicle placode formation
          platelet aggregation
          genetic imprinting
          regulation of parathyroid hormone secretion
          GTPase activity
          signal transducer activity
          insulin-like growth factor receptor binding
          protein binding
          GTP binding
          G-protein beta/gamma-subunit complex binding
          beta-2 adrenergic receptor binding
          D1 dopamine receptor binding
          mu-type opioid receptor binding
          ionotropic glutamate receptor binding
          metal ion binding
          corticotropin-releasing hormone receptor 1 binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-dd86746ff-lmgjf:80/100.66.77.199:80.
          git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.53.1-Offline