| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000456367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000456367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000456367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000456367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000456367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000456367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000456367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000456367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000456367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000614931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000614931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000614931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000614931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000614931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000617020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000617020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000617020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
ENST00000617020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007403.hg.1 |
ENST00000430861.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007403.hg.1 |
ENST00000430861.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377861.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000377865.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000456367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000456367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000456367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000456367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000456367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000456367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000456367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000456367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000456367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000544246.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000614931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000614931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000614931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000614931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000614931.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000617020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000617020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000617020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
ENST00000617020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_020403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009165.hg.1 |
NM_203487.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007403.hg.1 |
ENST00000430861.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007403.hg.1 |
ENST00000430861.1 |
Ensembl |