Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04181697_gH
          See other HNRNPCL2 GE Assays ›
          Gene Symbol:
          HNRNPCL2
          Gene Name:
          heterogeneous nuclear ribonucleoprotein C like 2
          Gene Aliases:
          HNRNPCP5
          Chromosome Location:
          Chr.1: 269828 - 271194 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04181697_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HNRNPCL2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001136561.3 NP_001130033.3 2 1046 93
          GenBank mRNA BC144551.1 - 2 1046 93
          BC157858.1 - 1 898 93
          BC171863.1 - 1 898 93

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Process(es)

          nucleic acid binding
          RNA binding
          protein binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CNV Microarray 2x400K 42056 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1896 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P136161 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15025867 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15025826 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15025864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00051477 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00051478 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20063684 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00051476 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20063649 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15025850 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20063679 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20063675 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15025847 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20063668 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20063655 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20063683 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15025854 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15025858 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20063663 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15025847 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20063649 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20063663 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00051476 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20063683 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20063684 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00051477 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20063679 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20063655 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00051478 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15025867 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15025858 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15025850 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20063668 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15025826 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15025864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20063675 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15025854 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00051472 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00051477 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00051475 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00051473 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15025850 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00051478 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00051468 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8047060 - -
          Affymetrix Human Gene 1.0 ST Array 7912553 - -
          Affymetrix Human Gene 1.0 ST Array 8047061 - -
          Affymetrix Human Gene 1.0 ST Array 7977660 - -
          Affymetrix Human Gene 1.0 ST Array 7977663 - -
          Affymetrix Human Gene 1.0 ST Array 7912581 - -
          Affymetrix Human Gene 1.0 ST Array 7977665 - -
          Affymetrix Human Gene 1.0 ST Array 7977668 - -
          Affymetrix Human Gene 1.0 ST Array 7977670 - -
          Affymetrix Human Gene 1.1 ST Array 7912580 - -
          Affymetrix Human Gene 1.1 ST Array 7912552 - -
          Affymetrix Human Genome U219 Array 11734649_x_at - -
          Affymetrix Human Genome U219 Array 11734649_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 216302_at - -
          Human Genome U133 Plus 2.0 Array 216302_PM_at - -
          Human Genome U133A 2.0 Array 216302_at - -
          Human Genome U133A 2.0 Array 216302_PM_at - -
          Human Genome U95 Set 32408_s_at - -
          Human Genome U95 Set 32408_PM_s_at - -
          Human Genome U133 Set 216302_at - -
          Human Genome U133 Set 216302_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A011317 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A011317 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3245015 NM_001136561.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3245015 BC144551.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3245015 BC157858.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3245015 BC171863.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3245015 BC137258.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3245015 NM_001136561.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3245015 BC144551.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3245015 BC157858.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3245015 BC171863.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3245015 BC137258.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3245015 NM_001136561.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3245015 BC144551.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3245015 BC157858.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3245015 BC171863.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3245015 BC137258.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3245015 NM_001136561.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3245015 BC144551.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3245015 BC157858.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3245015 BC171863.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3245015 BC137258.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0