| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8076494 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8076495 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8076496 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8076493 |
- |
- |
| Affymetrix Human Genome U219 Array |
11760598_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11760598_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012842.hg.1 |
ENST00000483050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014379.hg.1 |
ENST00000505920.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014379.hg.1 |
NM_001165877.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014379.hg.1 |
TC22000851.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008887.hg.1 |
ENST00000505920.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008887.hg.1 |
ENST00000505920.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008887.hg.1 |
ENST00000505920.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008887.hg.1 |
NM_001165877.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008887.hg.1 |
NM_001165877.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008887.hg.1 |
NM_001165877.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008887.hg.1 |
ENST00000505920.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008887.hg.1 |
ENST00000505920.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008887.hg.1 |
ENST00000505920.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008887.hg.1 |
NM_001165877.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008887.hg.1 |
NM_001165877.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008887.hg.1 |
NM_001165877.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012842.hg.1 |
ENST00000483050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164444.hg.1 |
ENST00000505920.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164444.hg.1 |
NM_001165877.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164444.hg.1 |
TC2200008887.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200164444.hg.1 |
ENST00000505920.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164444.hg.1 |
NM_001165877.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164445.hg.1 |
ENST00000505920.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164445.hg.1 |
NM_001165877.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164445.hg.1 |
TC2200008887.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200164445.hg.1 |
ENST00000505920.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164445.hg.1 |
NM_001165877.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164443.hg.1 |
ENST00000505920.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164443.hg.1 |
NM_001165877.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164443.hg.1 |
TC2200008887.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200164443.hg.1 |
ENST00000505920.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164443.hg.1 |
NM_001165877.1 |
Refseq |