Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04192464_mH
          See other HLA-DRB1 GE Assays ›
          Gene Symbol:
          HLA-DRB1
          Gene Name:
          major histocompatibility complex, class II, DR beta 1
          Gene Aliases:
          DRB1, DRw10, HLA-DR1B, HLA-DRB, SS1
          Chromosome Location:
          Chr.6: 32578769 - 32589836 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04192464_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HLA-DRB1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002124.3 NP_002115.2 2 - 3 460 103
          GenBank mRNA AK291987.1 - 2 - 3 460 103
          AY961072.1 - 2 - 3 366 103
          AY961073.1 - 2 - 3 366 103
          BC033827.1 - 2 - 3 409 5216243 103
          BC108922.1 - 2 - 3 40023604 103
          HG974574.1 - 2 - 3 366 103
          HG974596.1 - 2 - 3 366 103
          HG974600.1 - 2 - 3 366 103
          HM067861.1 - 2 - 3 366 103
          HM067862.1 - 2 - 3 366 103
          HM067863.1 - 2 - 3 366 103
          KJ534866.1 - 2 - 3 366 103
          L02545.1 - 2 - 3 380 103
          M16957.1 - 1 - 2 279 103
          M16959.1 - 1 - 2 279 103
          M17378.1 - 1 - 2 279 103
          M20430.1 - 2 - 3 409 103
          M20504.1 - 3 - 4 401 103
          M28583.1 - 3 - 4 405 103
          M28584.1 - 2 - 3 337 103
          U56640.1 - 1 - 2 223 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600014273.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06004123.hg.1, TC6_cox_hap2000193.hg.1, TC6_qbl_hap6000182.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HLA-DRB1

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          polysaccharide assembly with MHC class II protein complex
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          T cell costimulation
          T cell receptor signaling pathway
          interferon-gamma-mediated signaling pathway
          MHC class II protein complex binding
          peptide antigen binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_002124.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline