Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04194553_s1
          See other HTR5A GE Assays ›
          Gene Symbol:
          HTR5A
          Gene Name:
          5-hydroxytryptamine receptor 5A
          Gene Aliases:
          5-HT5A
          Chromosome Location:
          Chr.7: 155070324 - 155087392 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04194553_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HTR5A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024012.3 NP_076917.1 1 667 105
          GenBank mRNA AF498985.1 - 1 91 105
          AK094255.1 - 1 667 105
          BC112021.1 - 1 155 8327489 105
          BC113468.1 - 1 155 8322520 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700009796.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07001035.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HTR5A

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
          serotonin receptor signaling pathway
          cAMP-mediated signaling
          hippocampus development
          response to estradiol
          G-protein coupled serotonin receptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07016033.hg.1 TC07001035.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07016033.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07016033.hg.1 NM_024012.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009796.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009796.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009796.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009796.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009796.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009796.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009796.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009796.hg.1 ENST00000486819.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009796.hg.1 ENST00000486819.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009796.hg.1 ENST00000486819.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009796.hg.1 ENST00000486819.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009796.hg.1 NM_024012.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009796.hg.1 NM_024012.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009796.hg.1 NM_024012.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009796.hg.1 NM_024012.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009796.hg.1 NM_024012.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009796.hg.1 NM_024012.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009796.hg.1 NM_024012.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009796.hg.1 NM_024012.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07016035.hg.1 TC07001035.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07016035.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07016035.hg.1 ENST00000486819.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07016035.hg.1 NM_024012.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013163.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013163.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013163.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013163.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013163.hg.1 ENST00000493904.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013163.hg.1 ENST00000493904.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013163.hg.1 NR_038945.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013163.hg.1 NR_038945.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013163.hg.1 NR_038945.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013163.hg.1 NR_038945.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700169771.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169771.hg.1 NM_024012.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700169771.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169771.hg.1 NM_024012.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700169771.hg.1 TC0700009796.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0700013163.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013163.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013163.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013163.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013163.hg.1 ENST00000493904.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700013163.hg.1 ENST00000493904.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700013163.hg.1 NR_038945.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013163.hg.1 NR_038945.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013163.hg.1 NR_038945.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013163.hg.1 NR_038945.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700169770.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169770.hg.1 NM_024012.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700169770.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169770.hg.1 NM_024012.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700169770.hg.1 TC0700009796.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0700009796.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700009796.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700009796.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700009796.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700009796.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700009796.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700009796.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700009796.hg.1 ENST00000486819.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009796.hg.1 ENST00000486819.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009796.hg.1 ENST00000486819.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009796.hg.1 ENST00000486819.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009796.hg.1 NM_024012.3 Refseq
          Affymetrix Human Clariom D Assay TC0700009796.hg.1 NM_024012.3 Refseq
          Affymetrix Human Clariom D Assay TC0700009796.hg.1 NM_024012.3 Refseq
          Affymetrix Human Clariom D Assay TC0700009796.hg.1 NM_024012.3 Refseq
          Affymetrix Human Clariom D Assay TC0700009796.hg.1 NM_024012.3 Refseq
          Affymetrix Human Clariom D Assay TC0700009796.hg.1 NM_024012.3 Refseq
          Affymetrix Human Clariom D Assay TC0700009796.hg.1 NM_024012.3 Refseq
          Affymetrix Human Clariom D Assay TC0700009796.hg.1 NM_024012.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700169772.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169772.hg.1 NM_024012.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700169772.hg.1 ENST00000287907.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700169772.hg.1 NM_024012.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700169772.hg.1 TC0700009796.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline