Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04996411_g1
          See other CHMP5 GE Assays ›
          Gene Symbol:
          CHMP5
          Gene Name:
          charged multivesicular body protein 5
          Gene Aliases:
          C9orf83, CGI-34, HSPC177, PNAS-2, SNF7DC2, Vps60
          Chromosome Location:
          Chr.9: 33264879 - 33282069 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04996411_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CHMP5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001195536.1 NP_001182465.1 2 - 3 380 104
          NM_016410.5 NP_057494.3 2 - 3 380 104
          GenBank mRNA AF132968.1 - 2 - 3 211 104
          AF161525.1 - 2 - 3 200 104
          AK024753.1 - 2 - 3 228 104
          AK295744.1 - 2 - 3 220 104
          AK315455.1 - 2 - 3 229 104
          BC006974.1 - 2 - 3 210 3996277 104
          BC007457.1 - 2 - 3 242 4044464 104
          BC016698.1 - 2 - 3 259 4094210 104
          BC018603.1 - 2 - 3 221 104
          BC020796.1 - 2 - 3 211 4134310 104
          BC021168.1 - 2 - 3 225 3961351 104

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CHMP5

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell separation after cytokinesis
          regulation of receptor recycling
          nucleus organization
          lysosome organization
          mitotic metaphase plate congression
          endosome to lysosome transport
          regulation of centrosome duplication
          protein transport
          endosomal transport
          viral life cycle
          multivesicular body assembly
          viral budding
          multivesicular body sorting pathway
          cell-cell adhesion
          regulation of mitotic spindle assembly
          ESCRT III complex disassembly
          protein binding
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000487080.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000487080.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000487080.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 ENST00000487080.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017005.hg.1 ENST00000432760.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100017005.hg.1 ENST00000432760.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000223500.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000419016.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000487080.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000487080.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000487080.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 ENST00000487080.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_001195536.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_016410.5 Refseq
          Affymetrix Human Clariom D Assay TC0900006969.hg.1 NM_016410.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline