Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05042697_s1
          See other WNT10A GE Assays ›
          Gene Symbol:
          WNT10A
          Gene Name:
          Wnt family member 10A
          Gene Aliases:
          ECTD16, OODD, SSPS, STHAG4
          Chromosome Location:
          Chr.2: 218874116 - 218893928 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05042697_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: WNT10A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025216.3 NP_079492.2 4 1153 127
          XM_011511929.3 XP_011510231.1 5 1080 127
          XM_011511930.2 XP_011510232.1 3 773 127
          GenBank mRNA AB059569.1 - 4 1014 127
          AK024363.1 - 4 1472 127
          AK315081.1 - 4 1125 127
          AY009400.1 - 4 1205 127
          BC003544.1 - 2 637 127
          BC034352.1 - 4 1131 127
          BC040026.1 - 4 1442 127
          BC052234.1 - 4 1483 127

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene WNT10A

          Gene Ontology Categories:

          Function(s) Process(es)

          hair follicle development
          positive regulation of gene expression
          neural crest cell differentiation
          Wnt signaling pathway
          neuron differentiation
          hair follicle morphogenesis
          odontogenesis
          regulation of odontogenesis of dentin-containing tooth
          tongue development
          skin development
          cell fate commitment
          animal organ development
          epidermis morphogenesis
          sebaceous gland development
          canonical Wnt signaling pathway
          cellular response to transforming growth factor beta stimulus
          receptor binding
          frizzled binding
          cytokine activity
          receptor agonist activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 ENST00000258411.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 ENST00000258411.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 ENST00000258411.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 ENST00000258411.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 ENST00000258411.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 ENST00000258411.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 ENST00000258411.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 ENST00000258411.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 ENST00000258411.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 ENST00000258411.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 ENST00000458582.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 ENST00000458582.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 ENST00000483911.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 ENST00000483911.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 ENST00000489887.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 ENST00000489887.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 NM_025216.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 NM_025216.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 NM_025216.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 NM_025216.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 NM_025216.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 NM_025216.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 NM_025216.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 NM_025216.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010818.hg.1 NM_025216.2 Refseq
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 ENST00000258411.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 ENST00000258411.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 ENST00000258411.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 ENST00000258411.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 ENST00000258411.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 ENST00000258411.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 ENST00000258411.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 ENST00000258411.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 ENST00000258411.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 ENST00000258411.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 ENST00000458582.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 ENST00000458582.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 ENST00000483911.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 ENST00000483911.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 ENST00000489887.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 ENST00000489887.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 NM_025216.2 Refseq
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 NM_025216.2 Refseq
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 NM_025216.2 Refseq
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 NM_025216.2 Refseq
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 NM_025216.2 Refseq
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 NM_025216.2 Refseq
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 NM_025216.2 Refseq
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 NM_025216.2 Refseq
          Affymetrix Human Clariom D Assay TC0200010818.hg.1 NM_025216.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline