Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs05361842_g1
See other ZNF532 GE Assays ›
Gene Symbol:
ZNF532
LIMS4
Gene Name:
zinc finger protein 532
LIM zinc finger domain containing 4
Gene Aliases:
-
LIMS3L
Chromosome Location:
Chr.18: 58862924 - 58986480 on Build GRCh38
Chromosome Location:
Chr.2: 110359127 - 110473075 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID
Hs05361842_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ZNF532
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001318726.2
NP_001305655.1
3
1493
91
NM_001318727.2
NP_001305656.1
3
1454
91
NM_001318728.2
NP_001305657.1
4
1477
91
NM_001353525.2
NP_001340454.1
3
1518
91
NM_001353526.2
NP_001340455.1
4
1600
91
NM_001353527.2
NP_001340456.1
4
1442
91
NM_001353528.2
NP_001340457.1
3
1308
91
NM_001353529.2
NP_001340458.1
3
1360
91
NM_001353530.2
NP_001340459.1
4
1600
91
NM_001353531.2
NP_001340460.1
4
1503
91
NM_001353532.2
NP_001340461.1
3
1360
91
NM_001353533.2
NP_001340462.1
3
1493
91
NM_001353534.2
NP_001340463.1
3
1360
91
NM_001353535.2
NP_001340464.1
3
1421
91
NM_001353536.2
NP_001340465.1
3
1360
91
NM_001353537.2
NP_001340466.1
3
1360
91
NM_001353538.2
NP_001340467.1
3
1360
91
NM_001375912.1
NP_001362841.1
3
1759
91
NM_001375913.1
NP_001362842.1
3
1360
91
NM_018181.6
NP_060651.2
4
1575
91
NR_148459.2
4
1575
91
NR_148460.2
4
1536
91
XM_011526067.3
XP_011524369.1
1
1140
91
XM_017025809.2
XP_016881298.1
1
1140
91
XM_017025810.2
XP_016881299.1
6
1833
91
XM_017025811.3
XP_016881300.1
4
1957
91
XM_017025815.2
XP_016881304.1
2
1383
91
XM_017025823.2
XP_016881312.1
2
1384
91
XM_017025824.2
XP_016881313.1
2
1385
91
XM_017025829.2
XP_016881318.1
2
1384
91
XM_017025830.2
XP_016881319.1
2
1386
91
XM_047437581.1
XP_047293537.1
6
1858
91
XM_047437582.1
XP_047293538.1
5
1734
91
XM_047437583.1
XP_047293539.1
5
1640
91
XM_047437584.1
XP_047293540.1
5
1773
91
XM_047437585.1
XP_047293541.1
5
1701
91
XM_047437586.1
XP_047293542.1
4
1716
91
XM_047437587.1
XP_047293543.1
4
1619
91
XM_047437588.1
XP_047293544.1
4
1691
91
XM_047437589.1
XP_047293545.1
4
1652
91
XM_047437590.1
XP_047293546.1
4
1957
91
XM_047437591.1
XP_047293547.1
5
1798
91
XM_047437592.1
XP_047293548.1
5
1773
91
XM_047437593.1
XP_047293549.1
4
1691
91
XM_047437594.1
XP_047293550.1
4
1619
91
XM_047437595.1
XP_047293551.1
4
1981
91
XM_047437596.1
XP_047293552.1
5
1822
91
XM_047437597.1
XP_047293553.1
4
1582
91
XM_047437598.1
XP_047293554.1
4
1715
91
XM_047437599.1
XP_047293555.1
5
1797
91
XM_047437600.1
XP_047293556.1
2
1408
91
XM_047437601.1
XP_047293557.1
3
1435
91
XM_047437602.1
XP_047293558.1
1
1140
91
XM_047437603.1
XP_047293559.1
2
1383
91
XM_047437604.1
XP_047293560.1
1
1140
91
XM_047437605.1
XP_047293561.1
5
1798
91
XM_047437606.1
XP_047293562.1
5
1773
91
XM_047437607.1
XP_047293563.1
5
1773
91
XM_047437608.1
XP_047293564.1
2
1383
91
XM_047437609.1
XP_047293565.1
2
1388
91
GenBank mRNA
AB046849.2
-
3
1335
91
AF295077.1
-
4
1684
91
AK128058.1
-
4
1901
91
AY039256.1
-
3
2012
91
BC036366.1
-
4
1387
91
BC130618.1
-
3
1469
91
BC130620.1
-
4
1551
91
Gene Symbol: LIMS4
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_017003105.3
XP_016858594.1
10
25376
91
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length between 86 and 100
3' Most
Mammalian Gene Collection
Detect Genomic DNA
Homo sapiens
Both primers and probe map within a single exon
Matching Ambion siRNAs:
Click here to find siRNAs for Gene ZNF532
Click here to find siRNAs for Gene LIMS4
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
zinc ion binding
metal ion binding
protein binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline