Search
Search

NDUFB6| Gene Symbol: NDUFB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199987.2 | NP_001186916.1 | 3 | 655 | 102 | |
| NM_002493.5 | NP_002484.1 | 4 | 748 | 102 | ||
| NM_182739.3 | NP_877416.1 | 3 | 703 | 102 | ||
| GenBank mRNA | BG220924.1 | - | 2 | 384 | 102 | |
| BU952486.1 | - | 1 | 90 | 102 | ||
Set Membership: |
|