Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs99999034_m1
          See other CXCL8 GE Assays ›
          Gene Symbol:
          CXCL8
          Gene Name:
          C-X-C motif chemokine ligand 8
          Gene Aliases:
          GCP-1, GCP1, IL8, LECT, LUCT, LYNAP, MDNCF, MONAP, NAF, NAP-1, NAP1, SCYB8
          Chromosome Location:
          Chr.4: 73740569 - 73743716 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs99999034_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CXCL8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000584.4 NP_000575.1 1 - 2 152 81
          NM_001354840.3 NP_001341769.1 1 - 2 152 81
          GenBank mRNA AK131067.1 - 1 - 2 152 81
          AK311874.1 - 1 - 2 152 81
          BC013615.1 - 1 - 2 102 81
          BG497712.1 - 1 - 2 216 81
          BT007067.1 - 1 - 2 62 81
          CR542151.1 - 1 - 2 62 81
          DA671239.1 - 1 - 2 152 81
          M17017.1 - 1 - 2 152 81
          M26383.1 - 1 - 2 136 81
          Y00787.1 - 1 - 2 163 81
          Z11686.1 - 1 - 2 62 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400007836.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04000408.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Human Signal Transduction Pathways Human Cytokine Network HUMAN JAK-STAT PATHWAY HUMAN RHO FAMILY GTPASES HUMAN IL-6 PATHWAY 1700 Inventoried Human_Cytokine_Network Human_Signal_Transduction_Pathways Human_IL-6_Pathway Human_Rho_Family_GTPases Human_JAK-STAT_Pathway Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CXCL8

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          response to molecule of bacterial origin
          chemotaxis
          defense response
          inflammatory response
          immune response
          signal transduction
          G-protein coupled receptor signaling pathway
          negative regulation of cell proliferation
          positive regulation of gene expression
          negative regulation of gene expression
          calcium-mediated signaling
          regulation of cell adhesion
          neutrophil chemotaxis
          receptor internalization
          killing of cells of other organism
          response to endoplasmic reticulum stress
          intracellular signal transduction
          neutrophil activation
          cellular response to fibroblast growth factor stimulus
          regulation of single stranded viral RNA replication via double stranded DNA intermediate
          negative regulation of G-protein coupled receptor protein signaling pathway
          positive regulation of angiogenesis
          embryonic digestive tract development
          induction of positive chemotaxis
          negative regulation of cell adhesion molecule production
          antimicrobial humoral immune response mediated by antimicrobial peptide
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          positive regulation of neutrophil chemotaxis
          regulation of entry of bacterium into host cell
          cytokine activity
          interleukin-8 receptor binding
          protein binding
          chemokine activity
          heparin binding
          CXCR chemokine receptor binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000307407.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000307407.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000307407.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000307407.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000307407.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000307407.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000307407.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000307407.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000307407.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000307407.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000307407.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000401931.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000401931.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000401931.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000401931.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000401931.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000401931.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000401931.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000401931.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000483500.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000483500.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000483500.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000483500.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000483500.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 ENST00000483500.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 NM_000584.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 NM_000584.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 NM_000584.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 NM_000584.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 NM_000584.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 NM_000584.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 NM_000584.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 NM_000584.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 NM_000584.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 NM_000584.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007836.hg.1 NM_000584.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 176899 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline