Search
Search

MAD1L1| Gene Symbol: MAD1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549005.2 | XP_005549062.1 | 3 - 4 | 462 | 65 | |
| XM_005549006.2 | XP_005549063.1 | 3 - 4 | 462 | 65 | ||
| XM_015447005.1 | XP_015302491.1 | 3 - 4 | 462 | 65 | ||
| XM_015447006.1 | XP_015302492.1 | 3 - 4 | 416 | 65 | ||
| XM_015447007.1 | XP_015302493.1 | 3 - 4 | 1138 | 65 | ||
| XM_015447008.1 | XP_015302494.1 | 3 - 4 | 414 | 65 | ||
| XM_015447009.1 | XP_015302495.1 | 3 - 4 | 1125 | 65 | ||
| XM_015447010.1 | XP_015302496.1 | 3 - 4 | 414 | 65 | ||
| XM_015447011.1 | XP_015302497.1 | 3 - 4 | 462 | 65 | ||
| XM_015447013.1 | XP_015302499.1 | 3 - 4 | 463 | 65 | ||
| XM_015447014.1 | XP_015302500.1 | 3 - 4 | 463 | 65 | ||
Set Membership: |
|