Search
Search
| Gene Symbol: Bax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007527.3 | NP_031553.1 | 4 - 5 | 489 | 68 | |
| GenBank mRNA | AK013298.1 | - | 4 - 5 | 399 | 68 | |
| AK149994.1 | - | 4 - 5 | 382 | 68 | ||
| AK159233.1 | - | 4 - 5 | 454 | 68 | ||
| AY095934.1 | - | 3 - 4 | 836 | 68 | ||
| BC018228.1 | - | 4 - 5 | 434 | 3968903 | 68 | |
| BC053380.1 | - | 4 - 5 | 406 | 6489475 | 68 | |
| L22472.1 | - | 4 - 5 | 366 | 68 | ||
Set Membership: |
|