Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00436767_m1
          See other SPP1 GE Assays ›
          Gene Symbol:
          Spp1
          Gene Name:
          secreted phosphoprotein 1
          Gene Aliases:
          2AR, Apl-1, BNSP, BSPI, Bsp, ETA-1, Eta, OP, Opn, Opnl, Ric, Spp-1
          Chromosome Location:
          Chr.5: 104435111 - 104441053 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00436767_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001204201.1 NP_001191130.1 3 - 5 117
          NM_001204202.1 NP_001191131.1 3 - 4 249 114
          NM_001204203.1 NP_001191132.1 3 - 5 117
          NM_001204233.1 NP_001191162.1 2 - 3 294 114
          NM_009263.3 NP_033289.2 3 - 4 197 114
          GenBank mRNA AF515708.1 - 2 - 3 97 114
          AK133555.1 - 3 - 4 188 114
          AK145099.1 - 3 - 4 117
          AK149588.1 - 3 - 4 188 114
          AK150819.1 - 3 - 4 188 114
          AK151069.1 - 3 - 4 158 114
          AK151562.1 - 3 - 4 188 114
          AK153089.1 - 3 - 4 188 114
          AK154908.1 - 3 - 4 188 114
          AK160540.1 - 3 - 4 191 114
          AK165393.1 - 3 - 4 188 114
          AK165965.1 - 3 - 4 240 114
          AK166395.1 - 3 - 4 117
          AK171158.1 - 3 - 4 188 114
          BC002113.1 - 3 - 4 3489075 117
          BC020355.1 - 3 - 4 117
          BC029991.1 - 3 - 4 117
          BC057858.1 - 3 - 4 170 5294167 114
          BC080720.1 - 3 - 4 5252869 117
          CT010357.1 - 2 - 3 97 114
          J04806.1 - 3 - 4 166 114
          S78177.1 - 1 - 2 117
          X13986.1 - 2 - 3 112 114
          X16151.1 - 2 - 3 319 114

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001025.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spp1

          Panther Classification:

          Molecular Function -

          cytokine

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          osteoblast differentiation
          cell adhesion
          response to organic substance
          positive regulation of cell-substrate adhesion
          neutrophil chemotaxis
          biomineral tissue development
          response to vitamin D
          negative regulation of apoptotic process
          positive regulation of bone resorption
          response to steroid hormone
          cellular response to fluid shear stress
          cytokine activity
          extracellular matrix binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500008651.mm.1 TC0500001025.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500008651.mm.1 NM_001204201.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008651.mm.1 NM_001204201.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008651.mm.1 NM_001204202.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008651.mm.1 NM_001204202.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008651.mm.1 NM_001204203.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008651.mm.1 NM_001204203.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008651.mm.1 NM_001204233.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008651.mm.1 NM_001204233.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008651.mm.1 NM_009263.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008651.mm.1 NM_009263.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008652.mm.1 TC0500001025.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500008652.mm.1 NM_001204201.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008652.mm.1 NM_001204201.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008652.mm.1 NM_001204202.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008652.mm.1 NM_001204202.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008652.mm.1 NM_001204203.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008652.mm.1 NM_001204203.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008652.mm.1 NM_001204233.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008652.mm.1 NM_001204233.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008652.mm.1 NM_009263.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008652.mm.1 NM_009263.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008650.mm.1 TC0500001025.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500008650.mm.1 NM_001204201.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008650.mm.1 NM_001204201.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008650.mm.1 NM_001204202.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008650.mm.1 NM_001204202.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008650.mm.1 NM_001204203.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008650.mm.1 NM_001204203.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008650.mm.1 NM_001204233.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008650.mm.1 NM_001204233.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008650.mm.1 NM_009263.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008650.mm.1 NM_009263.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008653.mm.1 TC0500001025.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500008653.mm.1 NM_001204201.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008653.mm.1 NM_001204201.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008653.mm.1 NM_001204202.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008653.mm.1 NM_001204202.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008653.mm.1 NM_001204203.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008653.mm.1 NM_001204203.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008653.mm.1 NM_001204233.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008653.mm.1 NM_001204233.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008653.mm.1 NM_009263.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008653.mm.1 NM_009263.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 924589 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline