Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00439059_g1
          See other GAST GE Assays ›
          Gene Symbol:
          Gast
          Gene Name:
          gastrin
          Gene Aliases:
          GAS
          Chromosome Location:
          Chr.11: 100334412 - 100336996 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00439059_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gast
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010257.3 NP_034387.3 2 - 3 256 104
          GenBank mRNA AK008062.1 - 2 - 3 257 104
          AK008159.1 - 2 - 3 257 104
          AK008313.1 - 2 - 3 257 104
          AK008420.1 - 2 - 3 257 104
          AK008494.1 - 2 - 3 257 104
          AK008649.1 - 2 - 3 260 104
          BC064791.1 - 2 - 3 257 6431366 104

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gast

          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          hormone activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P236233 AK019058.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P236233 BC064791.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P236233 X94758.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P236233 NM_010257.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P236233 AK008062.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P236233 AK019058.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P236233 BC064791.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P236233 X94758.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P236233 NM_010257.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P236233 AK008062.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 38224 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 124736 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120511 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P236233 AK019058.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P236233 BC064791.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P236233 X94758.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P236233 NM_010257.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P236233 AK008062.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P236233 AK019058.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P236233 BC064791.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P236233 X94758.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P236233 NM_010257.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P236233 AK008062.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P236233 AK019058.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P236233 BC064791.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P236233 X94758.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P236233 NM_010257.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P236233 AK008062.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P236233 AK019058.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P236233 BC064791.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P236233 X94758.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P236233 NM_010257.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P236233 AK008062.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067493 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120511 - -
          Whole Mouse Genome G4122A A_52_P236233 AK019058.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P236233 BC064791.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P236233 X94758.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P236233 NM_010257.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P236233 AK008062.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P236233 AK019058.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P236233 BC064791.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P236233 X94758.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P236233 NM_010257.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P236233 AK008062.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136756 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120511 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09373316 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09373310 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09373302 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09373307 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09357460 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09373305 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09373311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09411077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09056747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09691013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09406662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09674885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09435230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09373315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09199705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09406664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09411076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09364199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09500595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09357463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09346326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09593317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09411078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09373313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09593319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09435229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09411080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09357462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09593316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28067455 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10381121 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381120 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381119 - -
          Affymetrix HT MG-430 PM Array Plate 1422915_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10381118 - -
          Mouse Genome 430 2.0 Array 1422915_at - -
          Mouse Genome 430 2.0 Array 1422915_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14459_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14459_PM_at - -
          Mouse Expression Set 430 1422915_PM_at - -
          Mouse Expression Set 430 1422915_at - -
          Murine Genome U74v2 Set 162472_PM_f_at - -
          Murine Genome U74v2 Set 98797_at - -
          Murine Genome U74v2 Set 98797_PM_at - -
          Murine Genome U74v2 Set 161639_f_at - -
          Murine Genome U74v2 Set 162472_f_at - -
          Murine Genome U74v2 Set 161639_PM_f_at - -
          Murine Genome U74v2 Set 169363_at - -
          Murine Genome U74v2 Set 169363_PM_at - -
          Mouse Genome 430A 2.0 Array 1422915_PM_at - -
          Mouse Genome 430A 2.0 Array 1422915_at - -
          Affymetrix HT Mouse Genome 430A Array 1422915_at - -
          Affymetrix HT Mouse Genome 430A Array 1422915_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 7000576 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007000576 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1241636 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241636 - -
          mouse-6 v1.1 (Illumina) scl40911.3.4_20-S - -
          Illumina Sentrix mouse-6.v1 bead array 7000576 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40911.3.4_20-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7000576 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1241636 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1241636 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline