Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00439389_m1
          See other HTR5B GE Assays ›
          Gene Symbol:
          Htr5b
          Gene Name:
          5-hydroxytryptamine (serotonin) receptor 5B
          Gene Aliases:
          5-Ht5b
          Chromosome Location:
          Chr.1: 121509758 - 121529312 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00439389_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Htr5b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010483.3 NP_034613.2 1 - 2 1051 76
          GenBank mRNA BC114988.1 - 1 - 2 940 40056207 76
          BC115825.1 - 1 - 2 940 40056204 76
          X69867.1 - 1 - 2 1086 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002973.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Htr5b

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          serotonin receptor signaling pathway
          signal transducer activity
          G-protein coupled receptor activity
          G-protein coupled serotonin receptor activity
          drug binding
          serotonin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20567330 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20567331 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20567327 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20567329 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P322115 X69867.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P322115 NM_010483.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P322115 X69867.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P322115 NM_010483.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354727 AK141560.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354727 BC036726.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354727 NM_153169.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354727 XM_001474312.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43664 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 29874 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 325106 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 775567 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 267667 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 776866 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 129902 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 153198 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 59993 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 91145 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120629 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136808 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P322115 X69867.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P322115 NM_010483.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P322115 X69867.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P322115 NM_010483.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P322115 X69867.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P322115 NM_010483.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P322115 X69867.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P322115 NM_010483.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20567342 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120629 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136808 - -
          Whole Mouse Genome G4122A A_51_P322115 X69867.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P322115 NM_010483.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P322115 X69867.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P322115 NM_010483.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110655 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125436 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120629 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136808 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00250921 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00250923 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00250922 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00250939 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00250928 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20567330 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20567331 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20567329 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20567327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20567346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00836535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00698987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00880941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01038270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01130163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01038268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00954504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01038266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00080852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00589528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00307744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00404712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00223146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01028777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00460052 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00856787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00589529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00250938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01029220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01029223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00191460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00261016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00250928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00954503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00856785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00900421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01029218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00926846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01130164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20567287 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10357291 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357289 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357290 - -
          Affymetrix HT MG-430 PM Array Plate 1422196_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10357288 - -
          Mouse Genome 430 2.0 Array 1422196_at - -
          Mouse Genome 430 2.0 Array 1422196_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15564_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15564_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100023949.mm.1 NM_010483.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023949.mm.1 NM_010483.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023949.mm.1 TC0100002973.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100023951.mm.1 NM_010483.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023951.mm.1 NM_010483.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023951.mm.1 TC0100002973.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100023953.mm.1 NM_010483.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023953.mm.1 NM_010483.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023953.mm.1 TC0100002973.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100023952.mm.1 NM_010483.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023952.mm.1 NM_010483.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023952.mm.1 TC0100002973.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100023950.mm.1 NM_010483.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023950.mm.1 NM_010483.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023950.mm.1 TC0100002973.mm.1 MTA TC
          Mouse Expression Set 430 1422196_PM_at - -
          Mouse Expression Set 430 1422196_at - -
          Murine Genome U74v2 Set 98810_at - -
          Murine Genome U74v2 Set 98810_PM_at - -
          Mouse Genome 430A 2.0 Array 1422196_PM_at - -
          Mouse Genome 430A 2.0 Array 1422196_at - -
          Affymetrix HT Mouse Genome 430A Array 1422196_at - -
          Affymetrix HT Mouse Genome 430A Array 1422196_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 516173 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 840594 - -
          Illumina mouse-6 v1.0 expression beadchip 840594 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000840594 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2595319 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2595319 - -
          mouse-6 v1.1 (Illumina) scl16367.3.1_212-S - -
          Illumina Sentrix mouse-6.v1 bead array 840594 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl16367.3.1_212-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 840594 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2595319 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2595319 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline