Search
Search
| Gene Symbol: Itk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281965.1 | NP_001268894.1 | 4 - 5 | 569 | 71 | |
| NM_001281966.1 | NP_001268895.1 | 4 - 5 | 551 | 71 | ||
| NM_001281967.1 | NP_001268896.1 | 4 - 5 | 551 | 71 | ||
| NM_001281968.1 | NP_001268897.1 | 4 - 5 | 548 | 71 | ||
| NM_010583.3 | NP_034713.2 | 4 - 5 | 551 | 71 | ||
| GenBank mRNA | AK079693.1 | - | 4 - 5 | 499 | 71 | |
| AK083380.1 | - | 4 - 5 | 522 | 71 | ||
| AK153613.1 | - | 4 - 5 | 498 | 71 | ||
| BC028478.1 | - | 4 - 5 | 493 | 71 | ||
| BC128374.1 | - | 4 - 5 | 477 | 40111687 | 71 | |
| BC128375.1 | - | 4 - 5 | 474 | 40111688 | 71 | |
| D14042.1 | - | 4 - 5 | 491 | 71 | ||
| L00619.1 | - | 4 - 5 | 556 | 71 | ||
| L05631.1 | - | 4 - 5 | 517 | 71 | ||
| L10628.1 | - | 4 - 5 | 519 | 71 | ||
Set Membership: |
|