Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00440280_g1
          See other MDK GE Assays ›
          Gene Symbol:
          Mdk
          Gene Name:
          midkine
          Gene Aliases:
          MK, Mek
          Chromosome Location:
          Chr.2: 91929805 - 91932297 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00440280_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mdk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012335.2 NP_001012335.1 2 - 3 250 62
          NM_001012336.2 NP_001012336.1 2 - 3 247 62
          NM_001291481.1 NP_001278410.1 2 - 3 157 62
          NM_001291483.1 NP_001278412.1 2 - 3 449 62
          NM_010784.5 NP_034914.1 2 - 3 449 62
          GenBank mRNA AK032742.1 - 2 - 3 153 63
          AK084457.1 - 2 - 3 170 64
          BC012244.1 - 2 - 3 118 4167496 62
          CF728526.1 - 2 - 3 143 62
          CF731708.1 - 2 - 3 246 63
          CF738400.1 - 2 - 3 255 70
          CJ174056.1 - 2 - 3 258 69
          DQ323887.1 - 2 - 3 420 62
          M19662.1 - 1 - 2 328 62
          M34327.1 - 2 - 3 123 62
          M34328.1 - 2 - 3 434 62
          M35833.1 - 2 - 3 123 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004129.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mdk

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          behavioral fear response
          multicellular organism development
          short-term memory
          response to wounding
          cell migration
          cerebellar granular layer development
          hippocampus development
          cell differentiation
          adrenal gland development
          defecation
          negative regulation of neuron apoptotic process
          positive regulation of transcription, DNA-templated
          regulation of behavior
          positive regulation of cell division
          negative regulation of neuron death
          growth factor activity
          heparin binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P107379 M34327.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P107379 M34328.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P107379 M35833.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P107379 DQ323886.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P107379 AY864926.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971897 M34327.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971897 M34328.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971897 M35833.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971897 DQ323886.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971897 DQ323887.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397019 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114776 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P107379 M34327.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P107379 M34328.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P107379 M35833.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P107379 DQ323886.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P107379 AY864926.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971897 M34327.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971897 M34328.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971897 M35833.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971897 DQ323886.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971897 DQ323887.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P453635 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P225665 M34327.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P225665 M34328.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P225665 M35833.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P225665 DQ323886.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P225665 DQ323887.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21425472 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114776 - -
          Whole Mouse Genome G4122A A_51_P225665 M34327.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225665 M34328.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225665 M35833.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225665 DQ323886.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225665 DQ323887.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453635 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112185 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114776 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01949036 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01949035 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01949031 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01202497 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01202498 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01949041 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01949042 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01949033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21425449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21425441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21425435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01949041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21425449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21425445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21425433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21425434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21425439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21425437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01949031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21425443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21425448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01949035 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10485075 - -
          Affymetrix Mouse Gene 1.0 ST Array 10485080 - -
          Affymetrix Mouse Gene 1.0 ST Array 10485071 - -
          Affymetrix Mouse Gene 1.0 ST Array 10485072 - -
          Affymetrix Mouse Gene 1.0 ST Array 10485078 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381602 - -
          Affymetrix Mouse Gene 1.0 ST Array 10485074 - -
          Affymetrix Mouse Gene 1.0 ST Array 10485077 - -
          Affymetrix Mouse Gene 1.0 ST Array 10485079 - -
          Affymetrix Mouse Gene 1.0 ST Array 10485073 - -
          Affymetrix Mouse Gene 1.0 ST Array 10485076 - -
          Affymetrix HT MG-430 PM Array Plate 1416006_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10485070 - -
          Affymetrix Mouse Gene 1.1 ST Array 10381601 - -
          Mouse Genome 430 2.0 Array 1416006_PM_at - -
          Mouse Genome 430 2.0 Array 1416006_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17242_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17242_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200034587.mm.1 NM_001012335.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034587.mm.1 TC0200004129.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200034584.mm.1 NM_001012335.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034584.mm.1 NM_001012336.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034584.mm.1 NM_010784.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034584.mm.1 TC0200004129.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200034592.mm.1 NM_001012336.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034592.mm.1 TC0200004129.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200034588.mm.1 NM_001012335.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034588.mm.1 TC0200004129.mm.1 MTA TC
          Mouse Expression Set 430 1416006_PM_at - -
          Mouse Expression Set 430 1416006_at - -
          Murine Genome U74v2 Set 160561_at - -
          Murine Genome U74v2 Set 160561_PM_at - -
          Mouse Genome 430A 2.0 Array 1416006_at - -
          Mouse Genome 430A 2.0 Array 1416006_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416006_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416006_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3840020 - -
          Illumina mouse-6 v1.0 expression beadchip 3840020 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003840020 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1254450 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254450 - -
          mouse-6 v1.1 (Illumina) scl18991.5.1_45-S - -
          Illumina Sentrix mouse-6.v1 bead array 3840020 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl18991.5.1_45-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3840020 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3158499 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1254450 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3079236 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3080012 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1254450 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3158499 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3080012 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3079236 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl18991.5.1_45-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline