Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00441538_m1
          See other SPA17 GE Assays ›
          Gene Symbol:
          Spa17
          Gene Name:
          sperm autoantigenic protein 17
          Gene Aliases:
          Sp17
          Chromosome Location:
          Chr.9: 37603294 - 37613833 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00441538_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spa17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011449.2 NP_035579.1 4 - 5 502 69
          GenBank mRNA AK005571.1 - 4 - 5 371 69
          BC059727.1 - 4 - 5 372 6775177 69
          BU936068.1 - 4 - 5 507 69
          CK137444.1 - 4 - 5 363 69
          Z46299.1 - 4 - 5 402 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002053.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spa17

          Gene Ontology Categories:

          Function(s) Process(es)

          binding of sperm to zona pellucida
          calmodulin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504065 NM_011449.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504065 AK005571.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504065 BC059727.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504065 Z46299.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504065 NM_011449.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504065 AK005571.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504065 BC059727.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504065 Z46299.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 576786 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 721684 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 709599 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418170 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 448705 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 458102 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 155289 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156673 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105354 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504065 NM_011449.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504065 AK005571.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504065 BC059727.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504065 Z46299.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504065 NM_011449.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504065 AK005571.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504065 BC059727.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504065 Z46299.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504065 NM_011449.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504065 AK005571.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504065 BC059727.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504065 Z46299.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504065 NM_011449.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504065 AK005571.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504065 BC059727.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504065 Z46299.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26431610 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156673 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105354 - -
          Whole Mouse Genome G4122A A_51_P504065 NM_011449.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504065 AK005571.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504065 BC059727.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504065 Z46299.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504065 NM_011449.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504065 AK005571.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504065 BC059727.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504065 Z46299.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124394 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156673 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105354 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08172362 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08172365 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08172364 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08172363 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07915677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26431634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07482741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07760302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07910594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07910593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08172365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08145133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08145127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07460118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26431560 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10592337 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592338 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592339 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592341 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592340 - -
          Affymetrix HT MG-430 PM Array Plate 1417635_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10592336 - -
          Mouse Genome 430 2.0 Array 1417635_at - -
          Mouse Genome 430 2.0 Array 1417635_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20686_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20686_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900017129.mm.1 NM_011449.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017129.mm.1 NM_011449.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017129.mm.1 TC0900002053.mm.1 MTA TC
          Mouse Expression Set 430 1417635_PM_at - -
          Mouse Expression Set 430 1417635_at - -
          Murine Genome U74v2 Set 101850_at - -
          Murine Genome U74v2 Set 101850_PM_at - -
          Mouse Genome 430A 2.0 Array 1417635_PM_at - -
          Mouse Genome 430A 2.0 Array 1417635_at - -
          Affymetrix HT Mouse Genome 430A Array 1417635_at - -
          Affymetrix HT Mouse Genome 430A Array 1417635_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 609169 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1190446 - -
          Illumina mouse-6 v1.0 expression beadchip 1190446 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001190446 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1249346 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249346 - -
          mouse-6 v1.1 (Illumina) scl36023.5.1_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 1190446 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36023.5.1_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1190446 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1249346 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1249346 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline