Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00442039_m1
          See other TNFRSF4 GE Assays ›
          Gene Symbol:
          Tnfrsf4
          Gene Name:
          tumor necrosis factor receptor superfamily, member 4
          Gene Aliases:
          ACT35, CD134, Ly-70, Ox40, TXGP1L, Txgp1
          Chromosome Location:
          Chr.4: 156013695 - 156016613 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00442039_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tnfrsf4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011659.2 NP_035789.1 4 - 5 608 72
          GenBank mRNA BC065782.1 - 4 - 5 533 72
          BC139239.1 - 4 - 5 478 8862261 72
          BC139266.1 - 4 - 5 478 8862288 72
          Z21674.1 - 4 - 5 429 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002103.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tnfrsf4

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          inflammatory response
          cellular defense response
          programmed cell death
          positive regulation of B cell proliferation
          tumor necrosis factor-mediated signaling pathway
          T cell proliferation
          regulation of apoptotic process
          negative regulation of sequence-specific DNA binding transcription factor activity
          regulation of protein kinase activity
          negative regulation of transcription, DNA-templated
          negative regulation of cytokine secretion
          positive regulation of immunoglobulin secretion
          negative regulation of activation-induced cell death of T cells
          negative regulation of extrinsic apoptotic signaling pathway
          tumor necrosis factor-activated receptor activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2125491 BC065782.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2125491 NM_011659.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 862377 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164728 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2125491 BC065782.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2125491 NM_011659.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P469433 BC065782.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P469433 NM_011659.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23441870 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164728 - -
          Whole Mouse Genome G4122A A_51_P469433 BC065782.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P469433 NM_011659.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123435 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164728 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03271335 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03271345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23441870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23441875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03271347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03961267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23441874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23441866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03271335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23441881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23441871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03271350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03271344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03961270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23441872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23441882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03271340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03961275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23441864 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10511289 - -
          Affymetrix Mouse Gene 1.0 ST Array 10511285 - -
          Affymetrix Mouse Gene 1.0 ST Array 10511283 - -
          Affymetrix Mouse Gene 1.0 ST Array 10511287 - -
          Affymetrix Mouse Gene 1.0 ST Array 10511288 - -
          Affymetrix Mouse Gene 1.0 ST Array 10511284 - -
          Affymetrix Mouse Gene 1.0 ST Array 10511286 - -
          Affymetrix HT MG-430 PM Array Plate 1420351_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10511282 - -
          Mouse Genome 430 2.0 Array 1420351_PM_at - -
          Mouse Genome 430 2.0 Array 1420351_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22163_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22163_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400017728.mm.1 NM_011659.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400017728.mm.1 NM_011659.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400017728.mm.1 TC0400002103.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400017727.mm.1 NM_011659.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400017727.mm.1 NM_011659.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400017727.mm.1 TC0400002103.mm.1 MTA TC
          Mouse Expression Set 430 1420351_PM_at - -
          Mouse Expression Set 430 1420351_at - -
          Murine Genome U74v2 Set 102681_PM_at - -
          Murine Genome U74v2 Set 102681_at - -
          Mouse Genome 430A 2.0 Array 1420351_at - -
          Mouse Genome 430A 2.0 Array 1420351_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420351_at - -
          Affymetrix HT Mouse Genome 430A Array 1420351_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 516191 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6840056 - -
          Illumina mouse-6 v1.0 expression beadchip 6840056 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006840056 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2446559 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2446559 - -
          mouse-6 v1.1 (Illumina) scl24600.8.1_49-S - -
          Illumina Sentrix mouse-6.v1 bead array 6840056 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24600.8.1_49-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6840056 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2446559 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2446559 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline