Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00442488_m1
          See other MAPK13 GE Assays ›
          Gene Symbol:
          Mapk13
          Gene Name:
          mitogen-activated protein kinase 13
          Gene Aliases:
          SAPK4, Serk4
          Chromosome Location:
          Chr.17: 28769317 - 28778704 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00442488_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mapk13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011950.2 NP_036080.2 3 - 4 406 65
          GenBank mRNA AK146993.1 - 3 - 4 394 65
          BC001992.1 - 3 - 4 375 3492903 65
          U81823.1 - 3 - 4 363 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000487.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mapk13

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein phosphorylation
          response to stress
          response to osmotic stress
          cell cycle
          phosphorylation
          peptidyl-serine phosphorylation
          positive regulation of interleukin-6 production
          intracellular signal transduction
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          MAP kinase activity
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31124215 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31124210 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31124214 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31124213 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31124209 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P239203 AK146993.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P239203 U81823.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P239203 NM_011950.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P239203 AK146993.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P239203 U81823.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P239203 NM_011950.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 657577 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 406952 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 646931 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 675472 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 896271 AF029748.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 24814 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166833 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P239203 AK146993.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P239203 U81823.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P239203 NM_011950.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P239203 AK146993.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P239203 U81823.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P239203 NM_011950.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P239203 AK146993.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P239203 U81823.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P239203 NM_011950.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P239203 AK146993.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P239203 U81823.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P239203 NM_011950.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31124194 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166833 - -
          Whole Mouse Genome G4122A A_51_P239203 AK146993.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P239203 U81823.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P239203 NM_011950.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P239203 AK146993.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P239203 U81823.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P239203 NM_011950.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130382 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166833 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13522219 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13522216 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13479290 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13247422 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31124215 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31124209 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31124213 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31124210 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31124214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31124295 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13065138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13522216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13479293 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124248 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13105903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13535882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13522218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13425516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13309206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13437878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13537634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13105904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13086433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13266363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13562704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13222268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13093798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31124181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13535879 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10443412 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443416 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443420 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443413 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443418 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443410 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443411 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443415 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443409 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443414 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443419 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443417 - -
          Affymetrix HT MG-430 PM Array Plate 1448871_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10443408 - -
          Mouse Genome 430 2.0 Array 1448871_PM_at - -
          Mouse Genome 430 2.0 Array 1448871_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26415_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26415_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700004230.mm.1 NM_011950.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004230.mm.1 NM_011950.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004230.mm.1 TC1700000487.mm.1 MTA TC
          Mouse Expression Set 430 1448871_at - -
          Mouse Expression Set 430 1448871_PM_at - -
          Murine Genome U74v2 Set 160763_at - -
          Murine Genome U74v2 Set 160763_PM_at - -
          Mouse Genome 430A 2.0 Array 1448871_at - -
          Mouse Genome 430A 2.0 Array 1448871_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448871_at - -
          Affymetrix HT Mouse Genome 430A Array 1448871_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 499918 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6760215 - -
          Illumina mouse-6 v1.0 expression beadchip 6760215 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006760215 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1212692 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1212692 - -
          mouse-6 v1.1 (Illumina) scl50917.9.3_9-S - -
          Illumina Sentrix mouse-6.v1 bead array 6760215 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50917.9.3_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6760215 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1212692 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2955452 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2955452 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1212692 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline