Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00444151_m1
          See other AK2 GE Assays ›
          Gene Symbol:
          Ak2
          Gene Name:
          adenylate kinase 2
          Gene Aliases:
          Ak-2, D4Ertd220e
          Chromosome Location:
          Chr.4: 128993217 - 129011529 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00444151_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ak2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033966.4 NP_001029138.1 2 - 3 342 61
          NM_016895.4 NP_058591.2 2 - 3 342 61
          GenBank mRNA AB020202.1 - 2 - 3 264 61
          AK010936.1 - 2 - 3 261 61
          AK010951.1 - 2 - 3 263 61
          AK050133.1 - 2 - 3 303 61
          AK077519.1 - 2 - 3 265 61
          AK166976.1 - 2 - 3 253 61
          AK167797.1 - 2 - 3 302 61
          AK168056.1 - 2 - 3 265 61
          AK168148.1 - 2 - 3 292 61
          BC008610.1 - 2 - 3 252 3584786 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001509.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ak2

          Panther Classification:

          Molecular Function -

          nucleotide kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleobase-containing compound metabolic process
          ADP biosynthetic process
          phosphorylation
          AMP metabolic process
          ATP metabolic process
          dATP metabolic process
          nucleotide phosphorylation
          nucleotide binding
          adenylate kinase activity
          ATP binding
          kinase activity
          transferase activity
          phosphotransferase activity, phosphate group as acceptor
          nucleobase-containing compound kinase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400011606.mm.1 NM_001033966.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011606.mm.1 NM_016895.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011606.mm.1 TC0400001509.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011608.mm.1 NM_001033966.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011608.mm.1 NM_016895.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011608.mm.1 TC0400001509.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011603.mm.1 NM_001033966.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011603.mm.1 NM_016895.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011603.mm.1 TC0400001509.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011604.mm.1 NM_001033966.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011604.mm.1 NM_016895.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011604.mm.1 TC0400001509.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011605.mm.1 NM_001033966.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011605.mm.1 NM_016895.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011605.mm.1 TC0400001509.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011602.mm.1 NM_001033966.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011602.mm.1 NM_016895.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011602.mm.1 TC0400001509.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 637459 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline