Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00445341_m1
          See other IL20 GE Assays ›
          Gene Symbol:
          Il20
          Gene Name:
          interleukin 20
          Gene Aliases:
          Zcyto10
          Chromosome Location:
          Chr.1: 130907192 - 130911296 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00445341_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il20
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021380.1 NP_067355.1 3 - 4 443 80
          GenBank mRNA AF224267.1 - 3 - 4 443 80
          AY876923.1 - 2 - 3 307 80
          BC111821.1 - 3 - 4 433 40049751 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003049.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Il20

          Gene Ontology Categories:

          Function(s) Process(es)

          inflammatory response
          immune response
          STAT protein import into nucleus
          hemopoiesis
          positive regulation of osteoclast differentiation
          positive regulation of JAK-STAT cascade
          receptor binding
          cytokine activity
          interleukin-20 receptor binding
          interleukin-22 receptor binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P331044 AK078698.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P331044 BC111821.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P331044 NM_021380.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P331044 AF224267.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P331044 AK078698.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P331044 BC111821.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P331044 NM_021380.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P331044 AF224267.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P132710 AK078698.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378023 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 911703 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 734578 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165249 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P331044 AK078698.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P331044 BC111821.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P331044 NM_021380.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P331044 AF224267.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P331044 AK078698.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P331044 BC111821.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P331044 NM_021380.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P331044 AF224267.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P331044 AK078698.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P331044 BC111821.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P331044 NM_021380.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P331044 AF224267.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P331044 AK078698.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P331044 BC111821.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P331044 NM_021380.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P331044 AF224267.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20612510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20612512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20612504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20612509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20612486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20612481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20612515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20612484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20612502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20612483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20612506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20612514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20612497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20612519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20612518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20612521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20612516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20612500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20612501 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165249 - -
          Whole Mouse Genome G4122A A_51_P331044 AK078698.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P331044 BC111821.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P331044 NM_021380.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P331044 AF224267.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P331044 AK078698.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P331044 BC111821.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P331044 NM_021380.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P331044 AF224267.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132710 AK078698.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104245 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165249 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01019961 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01019954 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01019951 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01019965 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01019959 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01019956 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01019955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20612451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612481 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612467 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00747318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00747320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00506021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00747316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00262312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01019961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00669076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00299047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01019951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01019959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612471 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612483 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20612517 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10357565 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357564 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357568 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357569 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357566 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357563 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357567 - -
          Affymetrix HT MG-430 PM Array Plate 1421608_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10357562 - -
          Mouse Genome 430 2.0 Array 1421608_at - -
          Mouse Genome 430 2.0 Array 1421608_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 58181_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 58181_at - -
          Affymetrix Mouse Clariom S Array PSR0100024469.mm.1 NM_021380.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100024469.mm.1 TC0100003049.mm.1 MTA TC
          Mouse Expression Set 430 1421608_at - -
          Mouse Expression Set 430 1421608_PM_at - -
          Mouse Genome 430A 2.0 Array 1421608_PM_at - -
          Mouse Genome 430A 2.0 Array 1421608_at - -
          Affymetrix HT Mouse Genome 430A Array 1421608_at - -
          Affymetrix HT Mouse Genome 430A Array 1421608_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4850647 - -
          Illumina mouse-6 v1.0 expression beadchip 4850647 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004850647 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230662 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230662 - -
          mouse-6 v1.1 (Illumina) scl16320.5.1_102-S - -
          Illumina Sentrix mouse-6.v1 bead array 4850647 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl16320.5.1_102-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4850647 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2925044 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2925044 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline