Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00445826_m1
          See other TNFRSF22 GE Assays ›
          Gene Symbol:
          Tnfrsf22
          Gene Name:
          tumor necrosis factor receptor superfamily, member 22
          Gene Aliases:
          2810028K06Rik, C130035G06Rik, SOBa, Tnfrh2, Tnfrsf1al2, mDcTrailr2
          Chromosome Location:
          Chr.7: 143634808 - 143649752 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00445826_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tnfrsf22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023680.3 NP_076169.2 1 - 2 115 114
          GenBank mRNA AJ278265.1 - 1 - 2 112
          AK012838.1 - 1 - 2 116 114
          AY046551.1 - 1 - 2 114 114
          AY165626.1 - 1 - 2 138 114
          AY165627.1 - 1 - 2 138 114
          BC076592.1 - 1 - 2 129 114

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004592.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AJ278265.1,AK012838.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tnfrsf22

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          receptor activity
          protein binding
          TRAIL binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155197 AY046551.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155197 AY165626.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155197 AY165627.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155197 BC076592.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155197 NM_023680.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P323852 AY046551.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P323852 BC076592.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P323852 NM_023680.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P323852 AK012838.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P323852 AY046551.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P323852 BC076592.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P323852 NM_023680.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P323852 AK012838.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 136705 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 921959 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 10253 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 50063 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 68650 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112663 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155197 AY046551.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155197 AY165626.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155197 AY165627.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155197 BC076592.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155197 NM_023680.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P323852 AY046551.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P323852 BC076592.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P323852 NM_023680.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P323852 AK012838.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P323852 AY046551.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P323852 BC076592.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P323852 NM_023680.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P323852 AK012838.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602210 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01659619 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06402282 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112663 - -
          Whole Mouse Genome G4122A A_52_P323852 AY046551.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P323852 BC076592.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P323852 NM_023680.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P323852 AK012838.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P323852 AY046551.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P323852 BC076592.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P323852 NM_023680.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P323852 AK012838.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134272 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135001 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112663 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06436300 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06112135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06437525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06436301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602195 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10569501 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569498 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569496 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569497 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569503 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569502 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569499 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569500 - -
          Affymetrix HT MG-430 PM Array Plate 1426095_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1422039_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1422038_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10569494 - -
          Mouse Genome 430 2.0 Array 1426095_PM_a_at - -
          Mouse Genome 430 2.0 Array 1426095_a_at - -
          Mouse Genome 430 2.0 Array 1422038_PM_a_at - -
          Mouse Genome 430 2.0 Array 1422038_a_at - -
          Mouse Genome 430 2.0 Array 1422039_PM_at - -
          Mouse Genome 430 2.0 Array 1422039_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 79202_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 79202_at - -
          Affymetrix Mouse Clariom S Array PSR0700038828.mm.1 NM_023680.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038828.mm.1 TC0700004592.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700038829.mm.1 NM_023680.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038829.mm.1 TC0700004592.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700038827.mm.1 NM_023680.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038827.mm.1 TC0700004592.mm.1 MTA TC
          Mouse Expression Set 430 1426095_a_at - -
          Mouse Expression Set 430 1422038_a_at - -
          Mouse Expression Set 430 1422038_PM_a_at - -
          Mouse Expression Set 430 1422039_at - -
          Mouse Expression Set 430 1422039_PM_at - -
          Mouse Expression Set 430 1426095_PM_a_at - -
          Mouse Expression Set 430 1442590_PM_at - -
          Mouse Expression Set 430 1442590_at - -
          Murine Genome U74v2 Set 129236_PM_at - -
          Murine Genome U74v2 Set 129236_at - -
          Mouse Genome 430A 2.0 Array 1426095_a_at - -
          Mouse Genome 430A 2.0 Array 1422039_PM_at - -
          Mouse Genome 430A 2.0 Array 1422038_a_at - -
          Mouse Genome 430A 2.0 Array 1422039_at - -
          Mouse Genome 430A 2.0 Array 1422038_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1426095_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1422039_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422039_at - -
          Affymetrix HT Mouse Genome 430A Array 1426095_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1422038_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1422038_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1426095_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 447194 - -
          ABI Mouse Genome Survey Microarray 469328 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 770400 - -
          MouseWG-6 v2 Expression BeadChip 105570722 - -
          MouseWG-6 v2 Expression BeadChip 5890731 - -
          Illumina mouse-6 v1.0 expression beadchip 5890731 - -
          Illumina mouse-6 v1.0 expression beadchip 5050050 - -
          Illumina mouse-6 v1.0 expression beadchip 770400 - -
          Illumina mouse-6 v1.0 expression beadchip 105570722 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005890731 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000770400 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005050050 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2454204 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1219749 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2467549 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2522818 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219749 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2467549 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2454204 - -
          mouse-6 v1.1 (Illumina) scl0319679.1_12-S - -
          mouse-6 v1.1 (Illumina) scl00319679.1_98-S - -
          mouse-6 v1.1 (Illumina) scl079202.1_17-S - -
          mouse-6 v1.1 (Illumina) scl0079202.2_327-S - -
          Illumina Sentrix mouse-6.v1 bead array 5890731 - -
          Illumina Sentrix mouse-6.v1 bead array 105570722 - -
          Illumina Sentrix mouse-6.v1 bead array 5050050 - -
          Illumina Sentrix mouse-6.v1 bead array 770400 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0319679.1_12-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0079202.2_327-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl079202.1_17-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 770400 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5050050 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105570722 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5890731 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2467549 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2454204 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2892413 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2467549 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2892413 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2454204 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2522818 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0079202.2_327-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline