Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00447994_m1
          See other NNMT GE Assays ›
          Gene Symbol:
          Nnmt
          Gene Name:
          nicotinamide N-methyltransferase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 48538001 - 48607635 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00447994_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nnmt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010924.2 NP_035054.1 2 - 3 478 73
          GenBank mRNA BC028757.1 - 2 - 3 394 1067421 73
          U86105.1 - 2 - 3 525 73
          U86106.1 - 2 - 3 525 73
          U86107.1 - 2 - 3 525 73
          U86108.1 - 2 - 3 525 73
          U89377.1 - 2 - 3 525 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002239.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nnmt

          Panther Classification:

          Molecular Function -

          methyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          methylation
          nicotinamide N-methyltransferase activity
          methyltransferase activity
          transferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2181557 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2185840 U86105.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2185840 U86106.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2185840 U86107.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2185840 U86108.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2185840 NM_010924.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 121064 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 496393 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 160383 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 395026 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 657075 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 486875 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 540542 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 525648 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 169282 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107455 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142912 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2181557 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2185840 U86105.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2185840 U86106.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2185840 U86107.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2185840 U86108.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2185840 NM_010924.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26492634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26492632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26492659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26492643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26492653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26492647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26492657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26492637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26492645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26492641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26492655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26492638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26492648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26492629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26492636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26492661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26492649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26492650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26492651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26492628 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06780234 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06780246 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107455 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142912 - -
          Whole Mouse Genome G4122A A_51_P476767 U86107.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476767 U86108.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126910 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107455 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142912 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07645851 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07459447 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07869250 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07459439 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07459438 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07459440 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07869246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26492556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07459445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07604291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07828065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07647574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07843662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07604299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07725257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07913022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08142200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07981235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07981240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07459439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07750437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07725260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07830798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07693477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26492661 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10593222 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593220 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593221 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593223 - -
          Affymetrix HT MG-430 PM Array Plate 1432517_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10593219 - -
          Mouse Genome 430 2.0 Array 1432517_a_at - -
          Mouse Genome 430 2.0 Array 1432517_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18113_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18113_at - -
          Affymetrix Mouse Clariom S Array PSR0900018674.mm.1 NM_010924.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018674.mm.1 TC0900002239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900018672.mm.1 NM_010924.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018672.mm.1 TC0900002239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900018673.mm.1 NM_010924.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018673.mm.1 TC0900002239.mm.1 MTA TC
          Mouse Expression Set 430 1432517_PM_a_at - -
          Mouse Expression Set 430 1432517_a_at - -
          Murine Genome U74v2 Set 101473_at - -
          Murine Genome U74v2 Set 101473_PM_at - -
          Mouse Genome 430A 2.0 Array 1432517_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1432517_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1432517_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1432517_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 651207 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104610286 - -
          MouseWG-6 v2 Expression BeadChip 450471 - -
          Illumina mouse-6 v1.0 expression beadchip 104610286 - -
          Illumina mouse-6 v1.0 expression beadchip 450471 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000450471 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2665409 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223025 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2665409 - -
          mouse-6 v1.1 (Illumina) scl35904.3.1_23-S - -
          Illumina Sentrix mouse-6.v1 bead array 104610286 - -
          Illumina Sentrix mouse-6.v1 bead array 450471 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35904.3.1_23-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104610286 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 450471 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2665411 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2885277 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2665411 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2885277 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2544305 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline