Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00448921_m1
          See other SYNGR3 GE Assays ›
          Gene Symbol:
          Syngr3
          Gene Name:
          synaptogyrin 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 24685092 - 24689949 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00448921_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Syngr3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011522.3 NP_035652.2 1 - 2 225 68
          GenBank mRNA AF117207.1 - 1 - 2 217 68
          AK048753.1 - 1 - 2 161 68
          AK080129.1 - 1 - 2 185 68
          AK081164.1 - 1 - 2 226 68
          AK082167.1 - 1 - 2 161 68
          AK087538.1 - 1 - 2 197 68
          BC025022.1 - 1 - 2 165 5369651 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001636.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Syngr3

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of transporter activity
          protein binding
          SH2 domain binding
          protein N-terminus binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099003 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099004 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099019 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099018 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099001 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099017 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099000 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099016 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403834 AK082167.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403834 AK087538.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403834 BC025022.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403834 AK080129.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403834 AK081164.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403834 AK082167.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403834 AK087538.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403834 BC025022.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403834 AK080129.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403834 AK081164.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 769760 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 403104 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 390882 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 40877 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P132178 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403834 AK082167.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403834 AK087538.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403834 BC025022.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403834 AK080129.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403834 AK081164.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403834 AK082167.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403834 AK087538.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403834 BC025022.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403834 AK080129.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403834 AK081164.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099027 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03293907 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P132178 - -
          Whole Mouse Genome G4122A A_51_P403834 AK082167.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403834 AK087538.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403834 BC025022.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403834 AK080129.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403834 AK081164.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403834 AK082167.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403834 AK087538.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403834 BC025022.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403834 AK080129.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403834 AK081164.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132192 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P132178 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13303167 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13303166 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13164830 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13303169 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13303163 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13303171 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099016 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099003 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099017 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099000 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099004 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099019 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099018 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31098995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31098986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31098990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31098992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13303167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31098995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13303169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31098996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31098984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13303164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13173904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13288896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31098981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31098982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13288908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31098987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13246119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31098977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13303163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31098989 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10448699 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448698 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448697 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448695 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448696 - -
          Affymetrix HT MG-430 PM Array Plate 1416098_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1448216_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10448694 - -
          Mouse Genome 430 2.0 Array 1416098_at - -
          Mouse Genome 430 2.0 Array 1448216_at - -
          Mouse Genome 430 2.0 Array 1448216_PM_at - -
          Mouse Genome 430 2.0 Array 1416098_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20974_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20974_at - -
          Affymetrix Mouse Clariom S Array PSR1700014645.mm.1 NM_011522.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014645.mm.1 NM_011522.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014645.mm.1 TC1700001636.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014647.mm.1 NM_011522.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014647.mm.1 NM_011522.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014647.mm.1 TC1700001636.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014648.mm.1 NM_011522.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014648.mm.1 NM_011522.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014648.mm.1 TC1700001636.mm.1 MTA TC
          Mouse Expression Set 430 1416098_PM_at - -
          Mouse Expression Set 430 1416098_at - -
          Mouse Expression Set 430 1448216_at - -
          Mouse Expression Set 430 1448216_PM_at - -
          Mouse Genome 430A 2.0 Array 1448216_PM_at - -
          Mouse Genome 430A 2.0 Array 1416098_at - -
          Mouse Genome 430A 2.0 Array 1448216_at - -
          Mouse Genome 430A 2.0 Array 1416098_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416098_at - -
          Affymetrix HT Mouse Genome 430A Array 1448216_at - -
          Affymetrix HT Mouse Genome 430A Array 1416098_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448216_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 750611 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4590576 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004590576 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2655862 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2655862 - -
          mouse-6 v1.1 (Illumina) scl50202.5_41-S - -
          Illumina Sentrix mouse-6.v1 bead array 4590576 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50202.5_41-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4590576 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2655862 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2655862 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl50202.5_41-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline