Search
Search
| Gene Symbol: Nes | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016701.3 | NP_057910.3 | 2 - 3 | 1023 | 72 | |
| GenBank mRNA | AF076623.2 | - | 2 - 3 | 987 | 72 | |
| AK009706.1 | - | 2 - 3 | 928 | 72 | ||
| AK031207.1 | - | 2 - 3 | 1049 | 72 | ||
| BC060693.1 | - | 2 - 3 | 1006 | 6410072 | 72 | |
| BC062893.1 | - | 2 - 3 | 1023 | 6405026 | 72 | |
Set Membership: |
|