Search
Search
| Gene Symbol: Ppt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002301.1 | - | 1 - 2 | 320 | 70 | |
| AK182405.1 | - | 1 - 2 | 29 | 70 | ||
| BC013462.1 | - | 1 - 2 | 319 | 4238225 | 70 | |
| BC052330.1 | - | 2 - 3 | 293 | 70 | ||
| CJ076607.1 | - | 2 - 3 | 278 | 70 | ||
| CX217389.1 | - | 2 - 3 | 256 | 70 | ||
| CX565661.1 | - | 2 - 3 | 251 | 70 | ||
Set Membership: |
|