Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00451586_m1
          See other MSMB GE Assays ›
          Gene Symbol:
          Msmb
          Gene Name:
          beta-microseminoprotein
          Gene Aliases:
          PIP, PSP94, beta-MSP
          Chromosome Location:
          Chr.14: 32142023 - 32158327 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00451586_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Msmb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020597.3 NP_065622.1 3 - 4 260 96
          GenBank mRNA U89840.1 - 2 - 3 228 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400000339.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Msmb

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P125636 U89840.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P125636 NM_020597.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P125636 U89840.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P125636 NM_020597.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 259080 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 170245 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 163990 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 818378 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 690 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 305763 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570051 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 802336 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 129164 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 65922 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 47141 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133136 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158252 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P125636 U89840.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P125636 NM_020597.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P125636 U89840.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P125636 NM_020597.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515864 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02826707 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133136 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08074633 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158252 - -
          Whole Mouse Genome G4122A A_52_P125636 U89840.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P125636 NM_020597.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P125636 U89840.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P125636 NM_020597.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134248 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133136 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11125571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11781984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11672599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11389510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11604469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11190846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11811490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11190843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515756 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11384847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11478172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11129879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11238280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11238274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11125572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11792286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11238283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515747 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11698470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11590521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11282858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11319041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515748 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515840 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10413838 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413837 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413836 - -
          Affymetrix HT MG-430 PM Array Plate 1421687_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10413835 - -
          Mouse Genome 430 2.0 Array 1421687_at - -
          Mouse Genome 430 2.0 Array 1421687_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17695_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17695_at - -
          Affymetrix Mouse Clariom S Array PSR1400002944.mm.1 NM_020597.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002944.mm.1 NM_020597.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002944.mm.1 TC1400000339.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400002945.mm.1 NM_020597.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002945.mm.1 NM_020597.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002945.mm.1 TC1400000339.mm.1 MTA TC
          Mouse Expression Set 430 1421687_PM_at - -
          Mouse Expression Set 430 1421687_at - -
          Murine Genome U74v2 Set 100715_at - -
          Murine Genome U74v2 Set 100715_PM_at - -
          Mouse Genome 430A 2.0 Array 1421687_PM_at - -
          Mouse Genome 430A 2.0 Array 1421687_at - -
          Affymetrix HT Mouse Genome 430A Array 1421687_at - -
          Affymetrix HT Mouse Genome 430A Array 1421687_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 415559 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4590070 - -
          Illumina mouse-6 v1.0 expression beadchip 4590070 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004590070 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1212724 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1212724 - -
          mouse-6 v1.1 (Illumina) scl46481.4.1_85-S - -
          Illumina Sentrix mouse-6.v1 bead array 4590070 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46481.4.1_85-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4590070 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1212724 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1212724 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline