Search
Search

Sh3kbp1| Gene Symbol: Sh3kbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135727.2 | NP_001129199.1 | 16 - 17 | 2313 | 98 | |
| NM_001135728.2 | NP_001129200.1 | 11 - 12 | 1315 | 98 | ||
| NM_001290661.1 | NP_001277590.1 | 5 - 6 | 995 | 98 | ||
| NM_001290664.1 | NP_001277593.1 | 4 - 5 | 866 | 98 | ||
| NM_021389.6 | NP_067364.2 | 15 - 16 | 1878 | 98 | ||
| NR_110969.1 | 15 - 16 | 1881 | 98 | |||
| NR_110970.1 | 15 - 16 | 1874 | 98 | |||
| GenBank mRNA | AK004636.1 | - | 15 - 16 | 1878 | 98 | |
| AK007283.1 | - | 5 - 6 | 981 | 98 | ||
| AK028299.1 | - | 11 - 12 | 1319 | 98 | ||
| AK030839.1 | - | 15 - 16 | 1882 | 98 | ||
| AK049182.1 | - | 16 - 17 | 2314 | 98 | ||
| AK052260.1 | - | 4 - 5 | 541 | 98 | ||
| AK080930.1 | - | 16 - 17 | 2179 | 98 | ||
| AK142295.1 | - | 5 - 6 | 769 | 98 | ||
| AK161512.1 | - | 5 - 6 | 996 | 98 | ||
| AK161735.1 | - | 11 - 12 | 1396 | 98 | ||
| AK172022.1 | - | 16 - 17 | 2294 | 98 | ||
| BC034646.1 | - | 15 - 16 | 1883 | 98 | ||
| BC052863.1 | - | 15 - 16 | 1823 | 98 | ||
Set Membership: |
|