Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00453659_m1
          See other RECQL4 GE Assays ›
          Gene Symbol:
          Recql4
          Gene Name:
          RecQ protein-like 4
          Gene Aliases:
          RecQ4
          Chromosome Location:
          Chr.15: 76703553 - 76710576 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00453659_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Recql4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_058214.3 NP_478121.2 8 - 9 1524 61
          GenBank mRNA AB039882.1 - 8 - 9 1397 61
          AB175741.1 - 8 - 9 1441 61
          AK090260.1 - 9 - 10 1128 61
          BC064680.1 - 8 - 9 1498 61
          BC131656.1 - 8 - 9 1437 61
          BC131657.1 - 7 - 8 1624 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001789.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Recql4

          Panther Classification:

          Molecular Function -

          DNA helicase

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA strand renaturation
          skeletal system development
          DNA replication
          DNA recombination
          positive regulation of cell proliferation
          DNA duplex unwinding
          pigmentation
          negative regulation of sister chromatid cohesion
          skeletal system morphogenesis
          nucleotide binding
          bubble DNA binding
          nucleic acid binding
          helicase activity
          ATP binding
          ATP-dependent helicase activity
          zinc ion binding
          hydrolase activity
          annealing helicase activity
          ATP-dependent 3'-5' DNA helicase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30350129 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30350130 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30350124 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30350126 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30350127 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30350125 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30350128 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2156016 BC064680.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2156016 NM_058214.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2156016 AK089539.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2156016 NM_027122.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086423 BC131656.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086423 BC064680.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086423 AK090260.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086423 AB175741.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086423 NM_058214.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 876915 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 684366 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 782321 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 49035 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101221 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086423 BC131656.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086423 BC064680.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086423 AK090260.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086423 AB175741.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086423 NM_058214.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2156016 BC064680.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2156016 NM_058214.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2156016 AK089539.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2156016 NM_027122.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P260178 BC131656.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P260178 BC131657.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P260178 BC064680.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P260178 AK090260.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P260178 AB175741.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350111 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101221 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08423173 - -
          Whole Mouse Genome G4122A A_51_P260178 BC131656.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P260178 BC131657.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P260178 BC064680.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P260178 AK090260.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P260178 AB175741.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102769 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101221 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12258141 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12258140 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12258134 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12258127 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30350128 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30350129 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30350130 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30350125 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30350126 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30350124 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30350127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30349996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30349998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12183660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12183635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12152379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12183617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12183643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12152382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12258136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12183616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350096 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10430092 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430077 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430076 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430081 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430086 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430094 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430089 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430084 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430079 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430080 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430088 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430090 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424999 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430075 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430072 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430087 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430085 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430083 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430078 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430082 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430074 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430073 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430091 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430093 - -
          Affymetrix HT MG-430 PM Array Plate 1422922_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10424991 - -
          Affymetrix Mouse Gene 1.1 ST Array 10430071 - -
          Mouse Genome 430 2.0 Array 1422922_at - -
          Mouse Genome 430 2.0 Array 1422922_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 79456_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 79456_at - -
          Affymetrix Mouse Clariom S Array PSR1500013877.mm.1 NM_058214.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013877.mm.1 NM_058214.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013877.mm.1 TC1500001789.mm.1 MTA TC
          Mouse Expression Set 430 1422922_at - -
          Mouse Expression Set 430 1422922_PM_at - -
          Mouse Genome 430A 2.0 Array 1422922_PM_at - -
          Mouse Genome 430A 2.0 Array 1422922_at - -
          Affymetrix HT Mouse Genome 430A Array 1422922_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422922_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 763321 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 540551 - -
          Illumina mouse-6 v1.0 expression beadchip 540551 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000540551 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2614945 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2614945 - -
          mouse-6 v1.1 (Illumina) scl47032.21.1_6-S - -
          Illumina Sentrix mouse-6.v1 bead array 540551 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl47032.21.1_6-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 540551 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3157773 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3078499 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3078499 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2614945 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3157773 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline