Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00454112_s1
          See other VMN1R229 GE Assays ›
          Gene Symbol:
          Vmn1r229
          Gene Name:
          vomeronasal 1 receptor 229
          Gene Aliases:
          V1re1
          Chromosome Location:
          Chr.17: 20814495 - 20815415 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00454112_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r229
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134190.1 NP_598951.1 1 276 112

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r229

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          response to pheromone
          pheromone binding
          pheromone receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P140855 NM_134190.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111583 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P140855 NM_134190.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P450432 NM_134190.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31083917 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111583 - -
          Whole Mouse Genome G4122A A_51_P450432 NM_134190.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140855 NM_134190.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111583 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13146641 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13146645 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13146637 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13146636 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13146634 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13404134 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13404128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31083928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13146644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13404128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13163705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31083948 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442150 - -
          Affymetrix Mouse Gene 1.1 ST Array 10442149 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 629583 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 110471 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000110471 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1224914 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224914 - -
          mouse-6 v1.1 (Illumina) scl8634.1.1_99-S - -
          Illumina Sentrix mouse-6.v1 bead array 110471 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl8634.1.1_99-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 110471 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1224914 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1224914 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline