Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00454883_m1
          See other PRDM15 GE Assays ›
          Gene Symbol:
          Prdm15
          Gene Name:
          PR domain containing 15
          Gene Aliases:
          C21orf83, E130018M06Rik, ORF62, Zfp298
          Chromosome Location:
          Chr.16: 97791467 - 97852427 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00454883_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prdm15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144789.2 NP_659038.2 21 - 22 2863 69
          GenBank mRNA AK144820.1 - 21 - 22 2768 69
          AY063457.1 - 2 - 3 421 69
          BC046433.1 - 2 - 3 422 69
          BC072597.1 - 19 - 20 2550 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600002157.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prdm15

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2173084 BC072597.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2173084 AK144820.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2173084 NM_144789.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P262118 BC046433.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P262118 BC072597.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P262118 AK144820.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P262118 AY063457.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P262118 AK053449.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P262118 BC046433.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P262118 BC072597.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P262118 AK144820.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P262118 AY063457.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P262118 AK053449.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 187649 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 448723 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 457656 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 720406 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 516101 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 118727 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 655204 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 690210 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 299883 AF033116.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 299883 AK032970.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 299883 AK122312.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 299883 AK141157.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 299883 BC060699.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 299883 NM_015753.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 125860 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 68659 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 751861 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 8533 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 736374 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 50901 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481319 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 619945 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 16084 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 183010 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 703011 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 265038 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630585 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 808120 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 118891 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 511898 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309572 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897057 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 940802 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 505959 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 758190 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 948902 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 489074 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 543782 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 477729 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309624 AF145044.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309624 AK150692.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309624 AK150827.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309624 AK167323.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309624 AY167925.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309624 BC004647.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309624 NM_008027.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309624 U90435.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654387 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 866748 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 90948 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 99665 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143391 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 155194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 7009 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154213 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137483 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2173084 BC072597.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2173084 AK144820.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2173084 NM_144789.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P262118 BC046433.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P262118 BC072597.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P262118 AK144820.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P262118 AY063457.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P262118 AK053449.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P262118 BC046433.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P262118 BC072597.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P262118 AK144820.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P262118 AY063457.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P262118 AK053449.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08699002 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137483 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154213 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08698891 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08699027 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08698909 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03267573 - -
          Whole Mouse Genome G4122A A_52_P262118 BC046433.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P262118 BC072597.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P262118 AK144820.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P262118 AY063457.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P262118 AK053449.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P262118 BC046433.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P262118 BC072597.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P262118 AK144820.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P262118 AY063457.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P262118 AK053449.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476900 AK040683.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476900 AK053449.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476900 NM_144789.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154213 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12732917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13043942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13020611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001399 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12968610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12616048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001373 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12739676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13043938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13043937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13043941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13020597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12589307 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12616050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12681525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31001396 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10441285 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441307 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441286 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441282 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441302 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441301 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441283 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441303 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441287 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441290 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441297 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441304 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441288 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441289 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441299 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441306 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441298 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441281 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441294 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441296 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441295 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441300 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441292 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441284 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441291 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441293 - -
          Affymetrix HT MG-430 PM Array Plate 1455459_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10441280 - -
          Mouse Genome 430 2.0 Array 1455459_PM_at - -
          Mouse Genome 430 2.0 Array 1455459_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 114604_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 114604_at - -
          Affymetrix Mouse Clariom S Array PSR1600016685.mm.1 NM_144789.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016685.mm.1 NM_144789.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600016685.mm.1 TC1600002157.mm.1 MTA TC
          Mouse Expression Set 430 1455459_at - -
          Mouse Expression Set 430 1455459_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455459_at - -
          Affymetrix HT Mouse Genome 430B Array 1455459_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2488164 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline