Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00458332_m1
          See other SARS2 GE Assays ›
          Gene Symbol:
          Sars2
          Gene Name:
          seryl-aminoacyl-tRNA synthetase 2
          Gene Aliases:
          2410015F05Rik, D7Ertd353e, SerRS, SerRSmt
          Chromosome Location:
          Chr.7: 28741934 - 28753879 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00458332_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sars2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023637.3 NP_076126.2 2 - 3 410 83
          GenBank mRNA AB029949.1 - 2 - 3 382 83
          AK010491.1 - 2 - 3 388 83
          AK087606.1 - 2 - 3 377 83
          AK140760.1 - 2 - 3 377 83
          BC079664.1 - 2 - 3 387 30357834 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000509.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sars2

          Panther Classification:

          Molecular Function -

          aminoacyl-tRNA synthetase translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          tRNA aminoacylation for protein translation
          seryl-tRNA aminoacylation
          nucleotide binding
          aminoacyl-tRNA ligase activity
          serine-tRNA ligase activity
          ATP binding
          ligase activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25023338 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25023337 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25023339 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462814 AK140760.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462814 BC079664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462814 AK010491.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462814 AK087606.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462814 NM_023637.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462814 AK140760.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462814 BC079664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462814 AK010491.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462814 AK087606.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462814 NM_023637.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 326992 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 737293 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 823250 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 402593 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 178003 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 835136 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 970671 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 49640 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462814 AK140760.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462814 BC079664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462814 AK010491.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462814 AK087606.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462814 NM_023637.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462814 AK140760.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462814 BC079664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462814 AK010491.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462814 AK087606.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462814 NM_023637.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P462814 AK140760.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P462814 BC079664.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P462814 AK010491.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P462814 AK087606.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P462814 NM_023637.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P462814 AK140760.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P462814 BC079664.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P462814 AK010491.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P462814 AK087606.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P462814 NM_023637.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25023362 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06150244 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01490512 - -
          Whole Mouse Genome G4122A A_51_P462814 AK140760.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P462814 BC079664.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P462814 AK010491.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P462814 AK087606.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P462814 NM_023637.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P462814 AK140760.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P462814 BC079664.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P462814 AK010491.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P462814 AK087606.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P462814 NM_023637.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103906 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01490507 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06461189 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06461180 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06461191 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06461193 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25023338 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25023337 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25023339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25023422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05972004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05929094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05929095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05971997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25023365 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10551563 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551564 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551561 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551565 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551555 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551567 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551569 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551566 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551557 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551560 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551556 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551562 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551558 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551559 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551568 - -
          Affymetrix HT MG-430 PM Array Plate 1448522_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10551554 - -
          Mouse Genome 430 2.0 Array 1448522_PM_at - -
          Mouse Genome 430 2.0 Array 1448522_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71984_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71984_at - -
          Affymetrix Mouse Clariom S Array PSR0700004146.mm.1 TC0700000509.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004146.mm.1 NM_023637.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004146.mm.1 NM_023637.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004147.mm.1 TC0700000509.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004147.mm.1 NM_023637.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004147.mm.1 NM_023637.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004149.mm.1 TC0700000509.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004149.mm.1 NM_023637.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004149.mm.1 NM_023637.3 Refseq
          Mouse Expression Set 430 1448522_at - -
          Mouse Expression Set 430 1448522_PM_at - -
          Murine Genome U74v2 Set 161339_f_at - -
          Murine Genome U74v2 Set 161339_PM_f_at - -
          Murine Genome U74v2 Set 161681_PM_i_at - -
          Murine Genome U74v2 Set 95630_at - -
          Murine Genome U74v2 Set 95630_PM_at - -
          Murine Genome U74v2 Set 161681_i_at - -
          Mouse Genome 430A 2.0 Array 1448522_at - -
          Mouse Genome 430A 2.0 Array 1448522_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448522_at - -
          Affymetrix HT Mouse Genome 430A Array 1448522_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 404424 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3162754 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3162754 - -
          mouse-6 v1.1 (Illumina) GI_12963764-I - -
          MouseRef-8 v2 Expression BeadChip ILMN_3162754 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3162754 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline