Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00459940_m1
          See other GPHA2 GE Assays ›
          Gene Symbol:
          Gpha2
          Gene Name:
          glycoprotein hormone alpha 2
          Gene Aliases:
          GPA2, zsig51
          Chromosome Location:
          Chr.19: 6226401 - 6227768 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00459940_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpha2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130453.3 NP_569720.1 3 - 4 322 75
          GenBank mRNA AF260740.1 - 3 - 4 322 75
          BC062804.1 - 3 - 4 322 1224990 75
          BC132274.1 - 2 - 3 310 40130551 75
          BC132562.1 - 2 - 3 310 40130839 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000106.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpha2

          Panther Classification:

          Molecular Function -

          peptide hormone intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein targeting to mitochondrion
          molecular_function
          hormone activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2100645 NM_130453.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2100645 AF260740.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2100645 BC062804.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 835373 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120949 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2100645 NM_130453.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2100645 AF260740.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2100645 BC062804.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P251833 NM_130453.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P251833 AF260740.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P251833 BC062804.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P251833 BC132274.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P251833 BC132562.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31933273 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120949 - -
          Whole Mouse Genome G4122A A_51_P251833 NM_130453.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P251833 AF260740.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P251833 BC062804.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P251833 BC132274.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P251833 BC132562.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123825 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125151 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120949 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14141627 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14141652 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14141646 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14141624 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14388658 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14388659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31933251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14260737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14144901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14144900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14141652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14141634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933272 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14148803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14388657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14186692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14141668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31933290 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10460786 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460785 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460784 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460783 - -
          Affymetrix HT MG-430 PM Array Plate 1421497_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10460782 - -
          Mouse Genome 430 2.0 Array 1421497_PM_at - -
          Mouse Genome 430 2.0 Array 1421497_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170458_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170458_at - -
          Affymetrix Mouse Clariom S Array PSR1900001225.mm.1 NM_130453.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001225.mm.1 NM_130453.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001225.mm.1 TC1900000106.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900001223.mm.1 NM_130453.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001223.mm.1 NM_130453.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001223.mm.1 TC1900000106.mm.1 MTA TC
          Mouse Expression Set 430 1421497_at - -
          Mouse Expression Set 430 1421497_PM_at - -
          Mouse Genome 430A 2.0 Array 1421497_PM_at - -
          Mouse Genome 430A 2.0 Array 1421497_at - -
          Affymetrix HT Mouse Genome 430A Array 1421497_at - -
          Affymetrix HT Mouse Genome 430A Array 1421497_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 643197 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6860070 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006860070 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245571 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245571 - -
          mouse-6 v1.1 (Illumina) scl52868.4.1_43-S - -
          Illumina Sentrix mouse-6.v1 bead array 6860070 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52868.4.1_43-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6860070 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1245571 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245571 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline